TLC Plates
EMD Millipore™ TLC Silica Gel 60 F254: 25 Glass plates, 20 x 20cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ TLC Silica Gel 60 F254: 100 Glass plates, 2.5 x 7.5cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ TLC Silica Gel 60 F254: 500 Glass plates, 2.5 x 7.5cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ Silica Gel 60 F254 Coated Aluminum-Backed TLC Sheets Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ TLC Plastic Sheets Available on GSA/VA Contract for Federal Government customers only.
Average particle size is 15μm with a 5-20μm distribution
EMD Millipore™ TLC Silica Gel 60 F254: 200 Glass plates, 5 x 10cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ TLC Silica Gel 60 F254: 50 Glass plates, 10 x 20cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
Selecto™ Flexible-Backed TLC Plates
Select from four media to match your separation needs
EMD Millipore™ TLC Silica Gel 60 F254: 100 Glass plates, 5 x 20cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ Precoated TLC and PLC Glass Plates Available on GSA/VA Contract for Federal Government customers only.
Average particle size is 15μm with a 5-20μm distribution
EMD Millipore™ PEI Cellulose F Coated TLC Sheets Available on GSA/VA Contract for Federal Government customers only.
For analysis of polar substances with exchange-active groups
EMD Millipore™ Aluminum Oxide 60 TLC Plates Available on GSA/VA Contract for Federal Government customers only.
For thin layer chromatography applications.
EMD Millipore™ Precoated Aluminum-Backed TLC Sheets Available on GSA/VA Contract for Federal Government customers only.
For analytical TLC or high-performance analytical TLC
EMD Millipore™ TLC LuxPlate™ Silica Gel 60 F254: 25 Glass plates, 20 x 20cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ Precoated HPTLC Glass Plates Available on GSA/VA Contract for Federal Government customers only.
Smaller size and distribution of HPTLC media yields improved resolution, shorter analysis times, and higher detection sensitivity
EMD Millipore™ Silica Gel 60G TLC Plates, Glass Backed Available on GSA/VA Contract for Federal Government customers only.
recommended for customer in QA/QC labs using older Ph Eur monograph methods, which require TLC plates with gypsum binder.
EMD Millipore™ TLC Silica Gel 60 Aluminum Sheets Available on GSA/VA Contract for Federal Government customers only.
For thin layer chromatography applications.
Kimble™ Chromatography TLC Plate Streaker
A simple device for uniformly streaking TLC plates.
EMD Millipore™ TLC and HPTLC GLP Plates, Glass Backed Available on GSA/VA Contract for Federal Government customers only.
For thin layer chromatography applications.
EMD Millipore™ TLC Silica Gel 60 F254: 25 Glass plates, 5 x 10cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ HPTLC LiChrospher Silica Gel 60 Plates/Sheets Available on GSA/VA Contract for Federal Government customers only.
thin layer chromatography plates based on spherical silica particles
EMD Millipore™ TLC Silica Gel 60, F254, MS-Grade Glass Plates, 20x20 Available on GSA/VA Contract for Federal Government customers only.
MS-Grade Glass TLC plates (20 x 20cm) with Silica gel 60 coated with fluorescent indicator F254, 25/Box
EMD Millipore™ Silica Gel 60 F254 TLC Plates Available on GSA/VA Contract for Federal Government customers only.
allow users to separate samples that vary greatly in size
EMD Millipore™ HPTLC Silica Gel 60 Aluminum Sheets Available on GSA/VA Contract for Federal Government customers only.
Aluminium HPTLC silica gel 60 plates. Silica gel HPTLC plates size 20 x 20 cm, 25 sheets.
EMD Millipore™ TLC Silica Gel 60 F254: 25 Glass plates, 5 x 20cm Available on GSA/VA Contract for Federal Government customers only.
Allows analyzation of any substance when combined with suitable mobile phase
EMD Millipore™ TLC Silica Gel 60, F254, Aluminum Plates, 10x10 Available on GSA/VA Contract for Federal Government customers only.
Aluminum TLC plates (10x10 cm) with Silica gel 60 coated with fluorescent indicator F254, 100/Box
DWK Life Sciences Kimble™ TLC Plate Streaker Base Assembly Only
Replacement Plate Streaker Base Assembly for use with Kimble™ 416430-0000.
  spinner