Stereo Microscopes
Laxco™ MZS4 Series Stereo Zoom Microscope
Laxco™ MZS4 Series revitalized old concept in stereo microscopy by incorporating high power LED to illuminate tilt able mirror. This creates a simple contrast method to see transparent specimens.
Laxco™ LMS-Z200 Series Stereo Zoom Microscope System
The LMS-Z200 series is ideal for routine laboratory applications. The combination of high quality optics, 6.4:1 zoom ratio, and advanced base/illumination options allow for complete flexibility of configuration.
Laxco™ LMS-S2 Series Fixed Magnification Stereo Microscope System
The LMS-S2 series features a fixed, dual magnification optical system, high-quality optics, and user-friendly controls ideal for entry level and academic applications. Multiple magnification, base, and illuminations options allow you to choose the configuration that best fits your needs.
Laxco™ MS Series Fixed Magnification Stereo Microscope System
Utilizes fixed dual magnification optical system. Laxco™ MS Series Fixed Magnification Stereo Microscope System are research-grade instrument with wide choice of zoom magnification ranges and most commonly found in general laboratory.
Walter Products Student Stereo 2X-4X Dual Illumination Microscope
Engineered for simplicity of operation, the ST Series stereo microscope provides a crisp three-dimensional erect image with a wide field of view. Simply change the magnification by turning the turret.
Fisherbrand™ Advanced Fixed Magnification Stereo Microscope - PROMO
Advanced Stereoscopic Microscope for increased capabilities in Macro-Size Specimens imaging
Laxco™ MZS1 Series Stereo Zoom Microscope
Most common instrument found in a laboratory. Laxco™ MZS1 Series Stereo Zoom Microscope combine stereoscopic zoom optical system with research grade optics.
Laxco™ T10 Series Track Stand Stereo Zoom Microscope
Large, low profile platform makes everyday work, simple and easy. Laxco™ T10 Series Track Stand Stereo Zoom Microscope can accommodate samples of all shapes and sizes easily and comfortably due to continuous vertical gear track.
Laxco™ MZS33 Series Stereo Microscope
Laxco™ MZ33 Series combines advanced contrast methods and simple LED fluorescence, and one of the common instrument found in laboratory.
Laxco™ LMS-Z200Pro Series Stereo Zoom Microscope System
The LMS-Z200Pro series features a 6.7:1 zoom ratio and research grade optics to provide a flat field of view, excellent depth of field and high resolution. The newly designed optical path with integrated click-stops delivers the precision required for life science research applications.
Laxco™ LMS-Z300 Series Stereo Zoom Microscope System
The LMS-Z300 is designed for research applications requiring higher magnification and resolution. It features and advanced optical design and 10:1 zoom ratio advanced applications.
Laxco™ D10 Series Stereo Zoom Microscope
Designed for wide range of applications. Laxco™ D10 Series Stereo Microscope has broad-range maginification head and its parfocal zoom control delivers crisp, clear images throughout zoom magnification range.
Laxco™ A10 Series Articulated Arm Stereo Zoom Microscope
Laxco™ A10 Series Articulated Arm Stereo Zoom Microscope design allows movement range for those applications where optics need to be moved over specimen in practically every conceivable position. Both vertical and horizontal reach all nearly any size object to be viewed.
Laxco™ LMS-Z100 Series Stereo Zoom Microscope System
Designed for the academic market, the LMS-Z100 series features a 4.4:1 zoom (8x-35x) and multiple base/illumination options for most applications. The small footprint, features, and economical price makes this a great fit for the classroom.
Laxco™ BM300 Series Boom Arm Stereo Zoom Microscope
Laxco™ BM300 Series Boom Arm Stereo Zoom Microscope has rugged industrial design that makes it perfect for harshest environment.
Laxco™ T40 Series Dual Track LED Stereo Zoom Microscope
Integrated cluster of 24 LEDs (720cd total) provide super bright white cooled illumination. Laxco™ T40 Series Dual Track LED Stereo Zoom Microscope can accommodate samples of all shapes and sizes easily and comfortably due to continuous vertical gear track.
Laxco™ LMS-Z400 Series Advanced Stereo Zoom Microscope System
The LMS-Z400 is the ideal scope for demanding life science research and laboratory applications. High-end features including high N.A. Plan Apochromatic optics, a 12.5:1 zoom ratio, and ergonomic tilting trinocular head all combine to offer both high resolution images and comfort.
Laxco™ MZS32 Series Stereo Microscope
Laxco™ MZS32 Series Stereo Microscope combines advanced contrast methods to deliver brilliant resolution for wide range of sample types. Its expandability to fluorescence makes this the ideal system for a growing labs needs.
Laxco™ MZS2 Series Pole Stand Stero Zoom Microscope
Most common instrument found in a laboratory. Laxco™ MZS2 Series Pole Stand Stereo Zoom Microscope combine stereoscopic zoom optical system with research grade optics.
Laxco™ BM 500 Series Stereo Zoom Microscope
Laxco™ BM500 Series utilizes large industrial base with both vertical and horizontal adjustments for imaging of large samples. Its dual heads allow for 2 users to view same field of view for surgical procedures or teaching.
Swift™ SM100 LED Stereo Microscopes Products for Science Education
Durable, versatile design is ideal for high school and college instruction.
Swift™ M29TZ-SM99CL-BTW1 Digital Screen Stereo Zoom Microscope Products for Science Education
Representing the latest in educational, laboratory and research technology, the Swift™ Optical Digital Screen Stereo Zoom Microscope can be used as a conventional stereo microscope or to share live images with your students or colleagues using WiFi tablets, WiFi enabled laptops and HD-ready LCD monitors or projectors through HDMI.
Ken-A-Vision™ Vision Scope 2 Stereo Microscopes Products for Science Education
High quality optics and rugged construction at a budget-friendly price.
Swift™ Stereo Trinocular Zoom Microscope Products for Science Education
Swift™ Optical SM105T-C Stereo Trinocular Zoom Microscope is a new addition to popular SM100 Stereo Series Microscopes, that features 10 to 30X zoom magnification (1x to 3x objectives).
Fisher Science Education™ Advanced Digital Stereomicroscopes Products for Science Education
Extra-rugged stereomicroscopes have a built-in high-resolution digital camera.
Celestron™ S10-60 Stereo Microscope Products for Science Education
Professional-level stereo microscope up to 60x power.
Fisher Science Education™ Intermediate Stereomicroscopes Products for Science Education
Stereomicroscopes feature rugged design and compact size.
Fisher Science Education™ Advanced StereoZoom Microscope Products for Science Education
An exceptional stereozoom microscope at an exceptionally low price.
Fisher Science Education™ Elementary Series Stereo Microscope Products for Science Education
Stereoscopic Microscope for Macro-Size Specimens is ideal for elementary school classrooms
Fisher Science Education™ Middle School Series Stereo Microscope Products for Science Education
Stereoscopic Microscope for Macro-Size Specimens is ideal for middle school classrooms
  spinner