Automated Urine Analyzers
BacterioScan™ 216Dx™ System for UTI Detection
Improved workflow reduces 60 to 80% of urine specimen plating
Germaine™ Laboratories AimStrip™ Urine Auto Analyzer
High volume urinalysis made easy! The AimStrip Urine Auto Analyzer is for the semi-quantitative detection of the following analytes in urine: Glucose, Bilirubin, Ketone (Acetoacetic acid), Specific Gravity, Blood, pH, Protein, Urobilinogen, and Leukocytes as well as the qualitative detection of Nitrite.
Germaine™ Laboratories AimStrip Urine Analyzer 2
A small and compact automated analyzer for the semi-quantitative detection of analytes in urine.
  spinner