Acids
Hydrochloric Acid, Certified ACS Plus, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Assay: 36.5 to 38.0% (w/w) Molecular Formula: HCl · H2O
Phosphotungstic Acid Hydrate (Crystalline/Certified), Fisher Chemical
Chemical Name or Material: Phosphotungstic Acid CAS: 12501-23-4 Purity Grade: Certified Grade Physical Form: Solid Molecular Formula: H3PO4 · 12 WO3 MDL Number: MFCD00149915 Synonym: Tungstophosphoric Acid
Acetic Acid, Glacial (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetic Acid, Glacial CAS: 64-19-7 Purity Grade: Certified ACS Grade Assay: ≥99.7% (w/w) Assay Percent Range: Assay: ≥99.7% w/w Molecular Formula: C2H4O2 Linear Formula: CH3COOH Formula Weight: 60.05 MDL Number: MFCD00036152
Phosphomolybdic Acid Hydrate (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Phosphomolybdic Acid Hydrate CAS: 51429-74-4 Purity Grade: Certified ACS Grade Physical Form: Solid Molecular Formula: H3MO12O40P Formula Weight: 1825.25 MDL Number: MFCD00149913 Synonym: Molybdophosphoric Acid Hydrate
Sulfuric Acid (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfuric Acid CAS: 7664-93-9 Purity Grade: TraceMetal Molecular Formula: H2SO4 Formula Weight: 98.08 MDL Number: MFCD00064589 Synonym: Oil of Vitriol
Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, ACROS Organics™
Chemical Name or Material: Acetic acid, sodium salt trihydrate CAS: 127-09-3 CAS Min %: 99.0 CAS Max %: 100.0 CAS: 6131-90-4 Purity Grade: ACS Reagent Grade Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C2H3NaO2·3H2O Linear Formula: CH3COONa·3H2O Formula Weight: 136.08 MDL Number: MFCD00071557 Synonym: Sodium acetate trihydrate
Acetic Acid, Glacial (USP/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetic Acid, Glacial CAS: 64-19-7 Purity Grade: FCC/USP Assay: 99.5 to 100.5% (USP) Molecular Formula: C2H4O2 Linear Formula: CH3COOH Formula Weight: 60.05 MDL Number: MFCD00036152 Synonym: Methanecarboxylic Acid
Hydrochloric Acid (Optima™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid CAS: 7647-01-0 Purity Grade: OPTIMA™ Assay: 32 to 35% (HCl, w/w) Molecular Formula: HCl · H2O Formula Weight: 36.46 MDL Number: MFCD00011324 Synonym: Muriatic Acid
Sulfuric Acid Concentrate (Certified) ACS, Fisher Chemical
Chemical Name or Material: Sulfuric Acid Concentrate CAS: 7664-93-9 CAS: 7732-18-5 Purity Grade: Certified Grade Physical Form: Liquid Molecular Formula: H2SO4 MDL Number: MFCD00064589 Synonym: Oil of Vitriol
Acetic Acid, Glacial (HPLC), Fisher Chemical
Chemical Name or Material: Acetic Acid, Glacial Name Note: 17.4N CAS: 64-19-7 Purity Grade: HPLC Grade Assay: ≥99.7% (w/w) Assay Percent Range: ≥99.7 w/w % Molecular Formula: C2H4O2 Linear Formula: CH3COOH Formula Weight: 60.05 MDL Number: MFCD00036152 Synonym: Methanecarboxylic Acid Purity Grade Notes: Also meets ACS specifications
Sulfurous Acid (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfurous Acid CAS: 7782-99-2 Purity Grade: Certified ACS Grade Assay: ≥6.0% Molecular Formula: H2SO3 MDL Number: MFCD00011335 Synonym: Sulfur Dioxide Solution
Nitric Acid (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Nitric Acid CAS: 7697-37-2 Purity Grade: TraceMetal Assay: 67 to 70% (HNO3, w/w) Molecular Formula: HNO3 Formula Weight: 63.01 MDL Number: MFCD00011349
Hydrochloric Acid (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid CAS: 7647-01-0 Purity Grade: TraceMetal Molecular Formula: HCl · H2O Formula Weight: 36.46 MDL Number: MFCD00011324 Synonym: Muriatic Acid
o-Phosphoric Acid, 85% (HPLC), Fisher Chemical
Chemical Name or Material: o-Phosphoric Acid Name Note: 85% CAS: 7664-38-2 CAS: 7732-18-5 Purity Grade: HPLC Grade Purity Grade Notes: Also meets ACS specifications Assay: ≥85.0% Assay Percent Range: ≥85.0 % Molecular Formula: H3O4P Formula Weight: 98 MDL Number: MFCD00011340
Nitric Acid (Certified ACS Plus), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Nitric Acid CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: Certified ACS Plus Grade Assay: 68.0 to 70.0% (w/w) Assay Percent Range: 70% Molecular Formula: HNO3 Formula Weight: 63.01 MDL Number: MFCD00011349
Hydrochloric Acid (NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric Acid CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: FCC/NF Assay: 36.5 to 38.0% (NF) Molecular Formula: HCl · H2O Formula Weight: 36.46 MDL Number: MFCD00011324 Synonym: Muriatic Acid
Formic Acid, 99.5+%, Optima™ LC/MS Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 64-18-6 Color: Colorless pH: 2.1 Physical Form: Liquid Molecular Formula: CH2O2
Nitric Acid (42° Be/Technical), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Nitric Acid Name Note: 42° Bé CAS: 7697-37-2 Purity Grade: Technical Grade Molecular Formula: HNO3 Formula Weight: 63.01 MDL Number: MFCD00011349
Sulfuric Acid (NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfuric Acid CAS: 7664-93-9 Purity Grade: FCC/NF Assay: 95.0 to 98.0% (FCC) Assay Percent Range: 95.0 to 98.0 % Molecular Formula: H2SO4 Formula Weight: 98.08 MDL Number: MFCD00064589 Synonym: Oil of Vitriol
Sulfuric Acid (Certified ACS Plus), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfuric Acid CAS: 7664-93-9 Purity Grade: Certified ACS Plus Grade Molecular Formula: H2SO4 Formula Weight: 98.08 MDL Number: MFCD00064589 UN Number: 1830
o-Phosphoric Acid, 85% (Certified ACS), Fisher Chemical
Chemical Name or Material: o-Phosphoric Acid Name Note: 85% CAS: 7664-38-2 CAS: 7732-18-5 Purity Grade: Certified ACS Grade Assay: ≥85.0% (w/w) Assay Percent Range: ≥85.0 w/w % Molecular Formula: H3O4P Formula Weight: 98 MDL Number: MFCD00011340 Synonym: Orthophosphoric Acid
Hydrochloric acid, ACS reagent, ca. 37% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrochloric acid Name Note: ca. 37% Solution in Water CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Assay: 36.5 to 38.0% Molecular Formula: ClH Linear Formula: HCl Formula Weight: 36.45 MDL Number: MFCD00011324 Synonym: Muriatic acid
Sulfurous Acid, ACS, 6.0% SO2 min, Alfa Aesar
Chemical Name or Material: Sulfurous acid Name Note: 6.0% SO2 min. CAS: 7782-99-2 Purity Grade: ACS Grade Physical Form: Liquid Molecular Formula: H2SO3 Formula Weight: 82.08 MDL Number: MFCD00011335 Synonym: Sulfur dioxide solution
Acetic acid, sodium salt, 99+%, extra pure, anhydrous, ACROS Organics™
Chemical Name or Material: Acetic acid, sodium salt Name Note: Anhydrous CAS: 127-09-3 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C2H3NaO2 Linear Formula: CH3CO2Na Formula Weight: 82.03 MDL Number: MFCD00012459
Acetic Acid, Glacial (TraceMetal™ Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetic Acid, Glacial CAS: 64-19-7 Purity Grade: TraceMetal Linear Formula: CH3COOH Formula Weight: 60.05 MDL Number: MFCD00036152 Synonym: Methanecarboxylic Acid, Vinegar Acid
Phosphomolybdic acid hydrate, ACS reagent, ACROS Organics™
Chemical Name or Material: Phosphomolybdic Acid hydrate CAS: 51429-74-4 CAS: 12026-57-2 Purity Grade: ACS Reagent Grade Physical Form: Crystals Molecular Formula: H3Mo12O40P·xH2O Linear Formula: H3[P(Mo3O10)4]·xH2O Formula Weight: 1825.25 MDL Number: MFCD00149913 Synonym: Molybdophosphoric Acid
Sulfuric Acid (Certified ACS Plus), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfuric Acid CAS: 7664-93-9 Purity Grade: Certified ACS Plus Grade Assay: 95.0 to 98.0% (w/w) Assay Percent Range: 95.0 to 98.0 w/w % Molecular Formula: H2SO4 Formula Weight: 98.08 MDL Number: MFCD00064589
Nitric Acid 67-69%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 7697-37-2 Certificate of Analysis: Certificate of Lot Analysis is included with each bottle. Chemical Name or Material: Nitric Acid Density: 1.40g/cm³ Boiling Point: 120.5°C Formula Weight: 63.01 Melting Point: −41°C Physical Form: Liquid Purity Grade: OPTIMA™ Merck Index: 15, 6663 Molecular Formula: HNO3 Synonym: Azotic Acid, Engraver's Acid
Acetic Acid, Glacial (USP/FCC/EP/BP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetic Acid, Glacial Name Note: 17.4N CAS: 64-19-7 Purity Grade: BP/EP/FCC/USP Assay: 99.0 to 100.5% (EP) Molecular Formula: C2H4O2 Linear Formula: CH3COOH Formula Weight: 60.05 MDL Number: MFCD00036152 Synonym: Methanecarboxylic Acid
Acetic Acid Solution, 1N (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetic Acid Solution, 1N CAS: 64-19-7 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: CH3CO2H Linear Formula: CH3COOH Formula Weight: 60.05 MDL Number: MFCD00036152 Synonym: Methanecarboxylic Acid
  spinner