Sulfurous Acid
Sulfurous Acid (Certified ACS), Fisher Chemical
CAS: 7782-99-2 Molecular Formula: H2O3S Molecular Weight (g/mol): 82.073 MDL Number: MFCD00011335 InChI Key: LSNNMFCWUKXFEE-UHFFFAOYSA-N Synonym: sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid PubChem CID: 1100 ChEBI: CHEBI:48854 IUPAC Name: sulfurous acid SMILES: OS(=O)O
Sulfurous Acid, ACS, 6.0% SO2 min, Alfa Aesar
CAS: 7782-99-2 Molecular Formula: H2O3S Molecular Weight (g/mol): 82.073 MDL Number: MFCD00011335 InChI Key: LSNNMFCWUKXFEE-UHFFFAOYSA-N Synonym: sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid PubChem CID: 1100 ChEBI: CHEBI:48854 IUPAC Name: sulfurous acid SMILES: OS(=O)O
Sulfurous Acid ACS AR, Macron Fine Chemicals™
CAS: 7782-99-2 Molecular Formula: H2O3S Molecular Weight (g/mol): 82.073 InChI Key: LSNNMFCWUKXFEE-UHFFFAOYSA-N Synonym: sulphurous acid, sulfur dioxide solution, schweflige saure german, h2so3, unii-j1p7893f4j, schweflige saure, acide sulfureux, acido sulfuroso, schweflige saeure, trioxosulfuric acid PubChem CID: 1100 ChEBI: CHEBI:48854 IUPAC Name: sulfurous acid SMILES: OS(=O)O
  spinner