Antibody Production and Purification Reagents and Kits
Molecular Probes™ Antibody Conjugate Purification Kit, 0.5-1 mg conjugate Non-distribution product as customer accommodation.
Antibody Conjugate Purification Kit, 0.5-1 mg conjugate
Gibson Bioscience Reagents: KOH 10%
For indicated test(s)
Gibson Bioscience Reagents: Oxidase Test
For the detection of oxidase producing organisms.
Gibson Bioscience Reagents: KOH 3%
Solubility test to aid in gram stain reactions
BD Cytometric Bead Array Mouse Th1/Th2 Cytokine Standard
Designed for use as a standard control with the Cytometric Bead Array (CBA) Mouse Th1/Th2 Cytokine Kit
Meridian Bioscience™ GullSORB™ IgG Inactivation Reagent
Diluted reagent for the inactivation of IgG antibodies in human serum
Gibson Bioscience Reagents: Ferric Chloride 10%
Used as a color developer for phenylalanine agar tests and tryptophan deaminase tests
Gibson Bioscience Reagents: Catalese
For the detection of microorganisms that possess the catalase enzyme
Gibson Bioscience Reagents: Kovacs
For the detection of microorganisms that produce Indole as a by-product of their metabolism
Gibson Bioscience Reagents: Alpha Napthol
Detect the production of acetoin, an intermediate in the formation of butylenes glycol, by organisms that ferment glucose by way of the butylenes glycol fermentation pathway
Gibson Bioscience Reagents: Nitrate A
For detection of the reduction of nitrate to nitrite
Gibson Bioscience Reagents: Nitrate B
For the detection of the reduction of nitrate to nitrite
MilliporeSigma™ Calbiochem™ TMB, Soluble Substrates for AP and HRP
Peroxidase substrate that produces blue soluble end product that can be read at 370nm or 650nm. Calbiochem™ TMB, Soluble Substrates for AP and HRP use of stop solution enhances sensitivity two- to four-fold and results in yellow solution that can be read at 450nm.
BD Cytometric Bead Array Human MIP-1α Standard
Useful as a standard control with the CBA Human MIP-1a Flex Set
BD Cytometric Bead Array Human Interleukin-13 Flex Set Standard
Reconstituted Human IL-13 Standard is useful as a standard control with the CBA Human IL-13 Flex Set
BD Cytometric Bead Array Mouse IL-4 Flex Set Standard
Standard control for flow cytometry
BD Cytometric Bead Array Human MCP-1 Standard
Useful as a standard control with the CBA Human MCP-1 Flex Set
BD Cytometric Bead Array Phospho JNK1/2 (T183/Y185) Standard
Lyophilized recombinant Phospho JNK1/2 (T183/Y185)
BD Cytometric Bead Array Mouse IFN-γ Flex Set Standard
Useful as a standard control with CBA Mouse IFN-γ Flex Set
BD Cytometric Bead Array Mouse IL-2 Flex Set Standard
Standard control for flow cytometry
  spinner