High Fidelity PCR Reagents and Kits
Thermo Scientific™ Phusion High-Fidelity DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Get superior performance for cloning and other applications requiring high fidelity with one of the most accurate thermostable DNA polymerases available.
Thermo Scientific™ Phusion™ Flash High-Fidelity PCR Master Mix Available on GSA/VA Contract for Federal Government customers only.
Perform short PCR protocols without compromising fidelity or yield with this master mix.
Invitrogen™ Platinum™ Taq DNA Polymerase High Fidelity Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ideal for amplification of DNA fragments when high yields and robust amplification are required
Invitrogen™ AccuPrime™ Taq DNA Polymerase, High Fidelity Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Amplifies nucleic acid templates using antibody-mediated hot-start, a blend of Taq DNA Polymerase and a proofreading enzyme, and AccuPrime accessory proteins for improved PCR fidelity, yield, and specificity over other hot-start DNA polymerases.
Thermo Scientific™ Phusion Site-Directed Mutagenesis Kit Available on GSA/VA Contract for Federal Government customers only.
Generate mutations with this fast and reliable system.
Thermo Scientific™ Phusion™ High-Fidelity DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
The most accurate thermostable DNA Polymerase available offering superior performance for cloning and other applications requiring high fidelity.
Thermo Scientific™ Phusion™ U Hot Start PCR Master Mix
Thermo Scientific Phusion U DNA Polymerase is a proofreading enzyme (25x Taq) with a proprietary mutation to read through uracils and incorporate dUTP
Thermo Scientific™ Phusion™ U Multiplex PCR Master Mix Available on GSA/VA Contract for Federal Government customers only.
High performance multiplex PCR master mix for fast and reliable simultaneous amplification of multiple targets
Thermo Scientific™ Phusion™ U Hot Start DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific Phusion U DNA Polymerase is a proofreading enzyme (25x Taq) with a proprietary mutation to read through uracils and incorporate dUTP
Thermo Scientific™ Phusion™ U Green Hot Start DNA Polymerase Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific Phusion U DNA Polymerase is a proofreading enzyme (25x Taq) with a proprietary mutation to read through uracils and incorporate dUTP
Thermo Scientific™ Phire™ Reaction Buffers
Buffers for use in PCR with Phire Hot Start II DNA Polymerase
Thermo Scientific™ Phusion™ U Multiplex PCR Master Mix Promotion
High performance multiplex PCR master mix for fast and reliable simultaneous amplification of multiple targets
  spinner