pH Reference Buffers
Buffer Solution, pH 4.00, Color-Coded Red (Certified), Fisher Chemical 
pH/temperature conversions given on label
Thermo Scientific™ Orion™ Standard All-in-One™ pH Buffer Kit
Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.
Buffer-Pac Color-Coded Solutions (Certified), Fisher Chemical 
CAS: 50-00-0 Molecular Formula: CH2O Molecular Weight (g/mol): 30.026 MDL Number: MFCD00163177 InChI Key: WSFSSNUMVMOOMR-UHFFFAOYSA-N PubChem CID: 712 ChEBI: CHEBI:16842 IUPAC Name: formaldehyde SMILES: C=O
Thermo Scientific™ Orion™ pH Buffer Bottles
Perform accurate, reproducible pH calibrations with color-coded Thermo Scientific™ Orion™ pH Buffer Bottles.
Buffer Solution, pH 7.00 (Certified), Fisher Chemical 
Potassium Phosphate Monobasic/Sodium Hydroxide
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready to use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches.
Buffer Solution, pH 8.00 (Certified), Fisher Chemical 
Potassium Phosphate Monobasic/Sodium Hydroxide
Buffer Solution, pH 9.00 (Certified), Fisher Chemical 
Boric Acid/Potassium Chloride/Sodium Hydroxide
Buffer Solution, pH 10.00 (Certified), Fisher Chemical 
Potassium Carbonate/Potassium Tetraborate/Potassium Hydroxide/Disodium EDTA Dihydrate
Thermo Scientific™ Orion™ pH Buffer Cubitainers™
Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.
Buffer Variety Pack, pH 4, 7, 10, Mettler Toledo™ 
Two 500mL bottle each of pH 4, 7, and 10 buffers
Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Sample well, rain, distilled/deionized, boiler feed and process waters with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor.
Thermo Scientific™ Orion™ ROSS™ All-in-One™ pH Buffer Kit
Maintain pH electrodes more effectively with Thermo Scientific™ Orion™ ROSS™ All-in-One™ pH Buffer Kits.
Buffer Solution, pH 7.40 (Certified), Fisher Chemical 
Potassium Phosphate Monobasic/Sodium Hydroxide