Wet Chemistry Buffers
Buffer Solution, TISAB, Fisher Chemical™ Available on GSA/VA Contract for Federal Government customers only.
For Analysis of the Determination of Fluoride Ion Concentration by Fluoride Ion Electrode
Ionic Strength Adjustment Low-Level TISAB II Buffer, SPEX CertiPrep™
Low Level TISAB II Ionic Strength Adjustment Buffer, 500 mL in H2O, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
  spinner