Inorganics Gold Compounds
Gold Chloride Trihydrate, Certified ACS, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 16961-25-4 Molecular Formula: HAuCl4 · 3 H2O MDL Number: MFCD00149904 Synonym: Gold(III) chloride trihydrate, Tetrachloroauric acid trihydrate, Hydrogren tetrachloroaurate(III), Tetrachloroauric(III) acid, Chloroauric acid trihydrate
Potassium tetrachloroaurate(III), 98%, ACROS Organics™
CAS: 13682-61-6 Molecular Formula: AuCl4K Molecular Weight (g/mol): 377.865 MDL Number: MFCD00043088 InChI Key: JMONCQYBAMKVQQ-UHFFFAOYSA-J Synonym: potassium tetrachloroaurate iii, potassium tetrachloroaurate, k aucl4, kaucl4 * 2h2o, potassium tetrachloridoaurate, potassium ion tetrachloroaurate, potassium tetrachloroaurate 1-, potassium tetrachloridoaurate 1-, potassium tetrachloridoaurate iii, potassium gold iii chloride PubChem CID: 11337898 ChEBI: CHEBI:51564 IUPAC Name: potassium;tetrachlorogold(1-) SMILES: Cl[Au-](Cl)(Cl)Cl.[K+]
Hydrogen tetrachloroaurate(III) trihydrate, ACS, 99.99% (metals basis), Au 49.5% min, Alfa Aesar™
CAS: 16961-25-4 Molecular Formula: HAuCl4·3H2O MDL Number: MFCD00149904 Synonym: Chloroauric acid
Gold(III) chloride, 99%, ACROS Organics™
CAS: 13453-07-1 Molecular Formula: AuCl3 MDL Number: MFCD00014172
Hydrogen tetrachloroaurate(III) hydrate, 99.999% (metals basis), Au 49% min, Alfa Aesar™ Premion™
CAS: 27988-77-8 Molecular Formula: AuCl4H3O Molecular Weight (g/mol): 357.79 MDL Number: MFCD00149903 InChI Key: OVJUDSKPNIBLOO-UHFFFAOYSA-K
Gold(III) acetate, 99.9% (metals basis), Alfa Aesar™
CAS: 15804-32-7 Molecular Formula: C6H9AuO6 Molecular Weight (g/mol): 374.099 MDL Number: MFCD00210614 InChI Key: OTCKNHQTLOBDDD-UHFFFAOYSA-K Synonym: gold 3+ triacetate, gold iii acetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1 PubChem CID: 15952561 IUPAC Name: gold(3+);triacetate SMILES: CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3]
Tetrachloroauric(III) Acid Trihydrate, 99.5%, For Analysis, EMSURE™, EMD Millipore™
CAS: 16961-25-4 Molecular Formula: AuCl4H·3H2O Synonym: Gold(III)chloride hydrochloric acid, Gold trichloride hydrochloride
Gold(III) chloride, Au 64.4% min, Alfa Aesar
CAS: 13453-07-1 Molecular Formula: AuCl3 MDL Number: MFCD00014172
Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Alfa Aesar™
CAS: 15283-45-1 Molecular Formula: AuH2Na3O6S4+ Molecular Weight (g/mol): 492.186 MDL Number: MFCD00046176 InChI Key: UCGZDNYYMDPSRK-UHFFFAOYSA-L Synonym: sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4 PubChem CID: 27176 SMILES: OS(=O)(=O)[S-].OS(=O)(=O)[S-].[Na+].[Na+].[Na+].[Au]
Gold(I) chloride, +99%, ACROS Organics™
CAS: 10294-29-8 Molecular Formula: AuCl Molecular Weight (g/mol): 232.417 MDL Number: MFCD00046175 InChI Key: FDWREHZXQUYJFJ-UHFFFAOYSA-M Synonym: gold i chloride, gold monochloride, gold chloride, aurous chloride, gold i chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade PubChem CID: 27366 ChEBI: CHEBI:30078 IUPAC Name: chlorogold SMILES: Cl[Au]
Gold(III) chloride, 99.99% (metals basis), Au 64.4% min, Alfa Aesar™, Premion™
CAS: 13453-07-1 Molecular Formula: AuCl3 MDL Number: MFCD00014172
Gold(I) cyanide, ACROS Organics™
CAS: 506-65-0 Molecular Formula: CAuN Molecular Weight (g/mol): 222.985 MDL Number: MFCD00003437 InChI Key: IZLAVFWQHMDDGK-UHFFFAOYSA-N Synonym: gold i cyanide, gold cyanide au cn, gold monocyanide, gold i cyanide, goldcarbonitrile, acmc-1cueu, gold i cyanide, premion r, gold i cyanide trace metals basis PubChem CID: 68172 IUPAC Name: gold(1+);cyanide SMILES: [C-]#N.[Au+]
  spinner