ISE Buffers
Buffer Solution (Acetate), pH 4.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Solution (Acetate), pH 4.01 CAS: 64-19-7 CAS: 127-09-3 CAS: 50-00-0 CAS: 67-56-1 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00137248 Synonym: Chlorine Titrimeter Reagent
Phosphate Buffer, pH 7, for Water Reaction in Aviation Fuel, Ricca Chemical
Chemical Name or Material: Phosphate Buffer Name Note: pH 7 CAS: 7732-18-5 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 7758-11-4 CAS Min %: 1.11 CAS Max %: 1.15 CAS: 7778-77-0 CAS Min %: 0.45 CAS Max %: 0.47 Physical Form: Liquid
Phosphate Buffer Solution, pH 6.8, for Determination of Phenols, Ricca Chemical
Chemical Name or Material: Phosphate Buffer Solution Name Note: pH 6.8 CAS: 7732-18-5 CAS Min %: 83.0 CAS Max %: 87.0 CAS: 7758-11-4 CAS Min %: 8.71 CAS Max %: 9.06 CAS: 7778-77-0 CAS Min %: 6.02 CAS Max %: 6.27 Physical Form: Liquid
Phosphate Buffer Solution, pH 6.8, APHA for Phenols, Ricca Chemical
Chemical Name or Material: Phosphate Buffer Solution
Ammonia ISE Fill Solution, Fisher Scientific™
Chemical Name or Material: Ammonia ISE Fill Solution CAS: 107-21-1 CAS: 7783-90-6 CAS: 12125-02-9 CAS: 7732-18-5 Physical Form: Liquid
Phosphate Buffer Solution, pH 7, for Chlorine Determination (ASTM Formulation), Ricca Chemical
Chemical Name or Material: Phosphate Buffer Solution Name Note: pH 7 CAS: 7732-18-5 CAS Min %: 92.0 CAS Max %: 96.0 CAS: 7558-79-4 CAS Min %: 3.14 CAS Max %: 3.27 CAS: 7778-77-0 CAS Min %: 2.33 CAS Max %: 2.43 CAS: 1310-73-2 CAS Min %: 0.34 CAS Max %: 0.36 Physical Form: Liquid
Acetate Buffer, pH 4.6, for Bromide Analysis, Ricca Chemical
Chemical Name or Material: Acetate Buffer Name Note: pH 4.6 CAS: 7732-18-5 CAS Min %: 82.0 CAS Max %: 86.0 CAS: 7647-14-5 CAS Min %: 7.42 CAS Max %: 7.72 CAS: 6131-90-4 CAS Min %: 5.61 CAS Max %: 5.83 CAS: 64-19-7 CAS Min %: 2.59 CAS Max %: 2.7 Physical Form: Liquid
Carbon Dioxide ISE Fill Solution, Fisher Scientific™
Chemical Name or Material: Carbon Dioxide ISE Fill Solution CAS: 107-21-1 CAS: 7783-90-6 CAS: 7647-14-5 CAS: 144-55-8 CAS: 7732-18-5 Physical Form: Liquid
Nitrate ISE Filling Solution, Fisher Scientific™
Chemical Name or Material: Nitrate ISE Filling Solution CAS: 7783-20-2 CAS: 7447-40-7 CAS: 7732-18-5 Physical Form: Liquid
Cyanide ISE Fill Solution, Fisher Scientific™
Chemical Name or Material: Cyanide ISE Fill Solution CAS: 7447-40-7 CAS: 7757-79-1 CAS: 7732-18-5 Physical Form: Liquid
Phosphate Buffer, for Cyanide, Ricca Chemical
Chemical Name or Material: Phosphate Buffer Name Note: For Cyanide CAS: 10049-21-5 CAS: 7732-18-5 Physical Form: Liquid
Buffer, for Fluoride, Citrate - Acetate, Ricca Chemical
Chemical Name or Material: Buffer CAS: 7732-18-5 CAS Min %: 85.0 CAS Max %: 89.0 CAS: 64-19-7 CAS Min %: 4.52 CAS Max %: 4.71 CAS: 7647-14-5 CAS Min %: 4.39 CAS Max %: 4.57 CAS: 1310-73-2 CAS Min %: 3.86 CAS Max %: 4.02 CAS: 6132-04-3 CAS Min %: 0.02 CAS Max %: 0.02 Physical Form: Liquid
Sörensen's Sodium Phosphate Buffer, 0.1 Molar Total Phosphate, pH 7.15, Ricca Chemical
Chemical Name or Material: Sörensen's Sodium Phosphate Buffer CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 7558-79-4 CAS Min %: 0.99 CAS Max %: 1.03 CAS: 10049-21-5 CAS Min %: 0.38 CAS Max %: 0.39 CAS: 89-83-8 CAS Min %: 0.01 CAS Max %: 0.01
Buffer Solution, for Nitrate Determination using Ion Selective Electrode, Ricca Chemical
Chemical Name or Material: Buffer Solution CAS: 7732-18-5 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 7784-31-8 CAS Min %: 1.66 CAS Max %: 1.73 CAS: 10294-26-5 CAS Min %: 0.33 CAS Max %: 0.34 CAS: 5329-14-6 CAS Min %: 0.24 CAS Max %: 0.25 CAS: 10043-35-3 CAS Min %: 0.12 CAS Max %: 0.13 CAS: 7732-18-5 Physical Form: Liquid
  spinner