Life Science Buffers
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents
Chemical Name or Material: Tris Base CAS: 77-86-1 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Powder/Solid Assay: ≥99.8% Assay Percent Range: ≥99.8 % Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 Synonym: Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents
Chemical Name or Material: Tris Hydrochloride CAS: 1185-53-1 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Solid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C4H11NO3 · HCl Linear Formula: H2NC(CH2OH)3·HCl Formula Weight: 157.6 Synonym: Tris(hydroxymethyl)aminomethane Hydrochloride
MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents
Chemical Name or Material: MOPS CAS: 1132-61-2 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Powder/Solid Assay: ≥97% Assay Percent Range: ≥97 % Molecular Formula: C7H15NO4S Linear Formula: CH2CH2OCH2CH2N(CH2)3SO3H Formula Weight: 209.26 Synonym: 3-(4-Morpholino)propane sulfonic acid
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Affymetrix/USB™
Chemical Name or Material: TE Buffer Physical Form: Solution Purity Grade: Molecular Biology Grade Concentration: 1X pH: 8 Packaging: Nalgene bottle
Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris Hydrochloride Name Note: 1M Solution, pH 8.0 CAS: 77-86-1 CAS: 1185-53-1 CAS: 7732-18-5 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: Tris-HCl
Tris Base (White Crystals or Crystalline Powder/Triple-Crystallized), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris Base Name Note: Alcohol-Free CAS: 77-86-1 Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Powder/Solid Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 Synonym: Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
Tris Hydrochloride, 1M Solution (pH 7.5/Mol. Biol.), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris Hydrochloride Name Note: 1M Solution, pH 7.5 CAS: 77-86-1 CAS: 1185-53-1 CAS: 7732-18-5 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: Tris-HCl
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical
Chemical Name or Material: Tris(hydroxymethyl)aminomethane CAS: 77-86-1 Purity Grade: Certified ACS Grade Physical Form: Powder/Solid Assay: 99.8 to 100.1% Assay Percent Range: 99.8 to 100.1% Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 MDL Number: MFCD00004679 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Trimethylol Aminomethane, Tris Buffer, Tris, Trizma™, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Bis-Tris, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Bis-Tris CAS: 6976-37-0 Physical Form: Powder/Solid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C8H19NO5 Linear Formula: (HOCH2CH2)2NC(CH2OH)3 Formula Weight: 209.24 Synonym: 2,2-Bis(hydroxymethyl)-2,2?,2?-nitrilotriethanol, 2, 2-Bis(hydroxymethyl)-2, 2?, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane
5X Kinase Buffer A Available on GSA/VA Contract for Federal Government customers only.
5X Kinase Buffer A
TR-FRET Dilution Buffer
TR-FRET Dilution Buffer
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris Hydrochloride Name Note: 1M Solution, pH 7.0 CAS: 77-86-1 CAS: 1185-53-1 CAS: 7732-18-5 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: Tris-HCl
HEPES, Free Acid, ULTROL™ Grade, Calbiochem™, EMD Millipore
Assay Percent Range: ≥99.0% Formula Weight: 238.3 Buffers: zwitterionic N-substituted aminosulfonic acid buffer CAS: 7365-45-9 Chemical Name or Material: HEPES, Free Acid Molecular Formula: C8H18N2O4S
Triethylammonium Acetate, 1 M Solution, Calbiochem™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Formula Weight: 161.2 CAS: 5204-74-0 Chemical Name or Material: Triethylammonium Acetate Concentration or Composition (by Analyte or Components): 1M Solution Molecular Formula: C8H19NO2
Imidazole, ULTROL™ Grade, Calbiochem™, EMD Millipore
Assay Percent Range: ≥99.0% Formula Weight: 68.1 CAS: 288-32-4 Chemical Name or Material: Imidazole Molecular Formula: C3H4N2
MOPS sodium salt, 98%, ACROS Organics™
Chemical Name or Material: MOPS sodium salt Name Note: 98% CAS: 71119-22-7 CAS Min %: 97.5 CAS Max %: 100.0 Assay: 98% Assay Percent Range: 98% Molecular Formula: C7H14NNaO4S Formula Weight: 231.25 MDL Number: MFCD00064350 Synonym: 3-(4-Morpholino)propanesulfonic acid sodium salt
MOPS, 99%, for biochemistry, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: MOPS Name Note: 99% CAS: 1132-61-2 CAS Min %: 98.5 CAS Max %: 100.0 Purity Grade: For biochemistry Assay: 99% Assay Percent Range: 99% Molecular Formula: C7H15NO4S Formula Weight: 209.26 MDL Number: MFCD00006183 Synonym: 3-(N-Morpholino)propanesulfonic acid
Tris (Tromethamine) (Granular/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris (Tromethamine) CAS: 77-86-1 Purity Grade: USP Physical Form: Powder/Solid Assay: 99.0 to 101.0% (USP) Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14 MDL Number: MFCD00004679 Synonym: Trishydroxymethylaminomethane, Tris Buffer, Tris, THAM, Trizma™, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Glycine, ACS, 98.5+%, Affymetrix/USB™
CAS: 56-40-6 Chemical Name or Material: Glycine Formula Weight: 75.07 Linear Formula: H2NCH2COOH MDL Number: MFCD00008131 Melting Point: ∽245°C (dec.) Molecular Formula: C2H5NO2 Physical Form: Crystalline Powder Purity Grade: ACS Grade Specific Gravity: 1.595
Tris, 99.8 to 100.1% (Dry Basis), Molecular Biology Grade, Ultrapure, Affymetrix/USB™
Chemical Name or Material: Tris Formula Weight: 121.14 Purity Grade: Molecular Biology Grade Physical Form: Crystalline Powder Linear Formula: NH2C(CH2OH)3 MDL Number: MFCD00004679 Molecular Formula: C4H11NO3 Synonym: Tris(hydroxymethyl)amino methane Boiling Point: 219° to 220°C Melting Point: 171° to 172°C Packaging: Plastic bottle
  spinner