Analytical n- Hexane
n-Hexane, 99+%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane CAS: 110-54-3 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, Anhydrous, Alfa Aesar
Chemical Name or Material: n-Hexane CAS: 110-54-3 Physical Form: Liquid Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD00009520
n-Hexane, 96+%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Hexane CAS: 110-54-3 CAS Min %: 96.0 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥96% Assay Percent Range: 96+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
Alfa Aesar™ n-Hexane, anhydrous, over molecular sieves, packaged under argon in resealable ChemSeal™ bottles
Chemical Name or Material: n-Hexane CAS: 110-54-3 Name Note: Anhydrous, over molecular sieves Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 UN Number: UN1208 Beilstein: 1730733 Merck Index: 14,4694
n-Hexane, 95+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane Name Note: 95% CAS: 110-54-3 CAS Min %: 95.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥95% Assay Percent Range: 95+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
Hexanes (95% n-hexane), ULTRA RESI-ANALYZED, J.T.Baker™
Chemical Name or Material: Hexanes (95% n-hexane) CAS: 110-54-3 Purity Grade: 99.5 % min. Assay Percent Range: 95% n-hexane Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18g/mol
Alfa Aesar™ n-Hexane, 99%
Chemical Name or Material: n-Hexane CAS: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 UN Number: UN1208 Beilstein: 1730733 Merck Index: 14,4694
Alfa Aesar™ Hexanes, mixed isomers, ACS, 98.5+%
Chemical Name or Material: Hexanes CAS: 92112-69-1 Name Note: Mixed isomers Purity Grade: ACS Grade Formula Weight: 86.18 MDL Number: MFCD02179311 UN Number: UN1208
Hexane, B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson
CAS Max %: <40.0000% Density: 0.659 - 0.673 g/cm3 Boiling Point: 68.7°C Formula Weight: 86.18 CAS Min %: >60.0000% Purity Grade: for HPLC, gradient grade CAS: 110-54-3 Flash Point: −26°C Chemical Name or Material: Hexane CAS Max %: >60.00% Beilstein: 1730733 Name Note: B&J Brand™, for HPLC, gradient grade, GC, pesticide residue analysis and spectrophotometry, >99.9% (>60% n-Hexane) Packaging: Glass Bottle Assay: >99.9% UN Number: UN1208
Hexane, ReagentPlus™, ≥99%, Honeywell™
Solubility: Insoluble in water Vapor Pressure: 5.2psi (37.7°C), 132mmHg (20°C), 256mmHg (37.7°C) Vapor Density: 3 (vs air) Density: 0.659g/mL at 25°C Explosion Limit: 0.077 Flash Point: −26°C (−14.8°F) Ignition Point: 453°F Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 UN Number: UN1208 Assay: ≥99% Formula Weight: 86.18g/mol Purity Grade: ReagentPlus™ Chemical Name or Material: Hexane CAS: 110-54-3 Beilstein: 1730733 Refractive Index: n20/D 1.375(lit.) Melting Point: −95°C Boiling Point: 66° to 69°C Color: Colorless Physical State: Liquid
n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane Name Note: Over Molecular Sieves CAS: 110-54-3 CAS Min %: 96.0 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: 97% Assay Percent Range: 97% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, 99% , 99.0% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane CAS: 110-54-3 Physical Form: Liquid Assay Percent Range: 99.0%, min. Molecular Formula: CH3(CH2)4CH3 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18
Alfa Aesar™ Hexanes, mixed isomers, (60+% n-hexane)
Chemical Name or Material: Hexanes CAS: 92112-69-1 Name Note: Mixed isomers Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD02179311 UN Number: UN1208
Alfa Aesar™ Hexanes, mixed isomers, Environmental Grade
Chemical Name or Material: Hexanes CAS: 92112-69-1 Name Note: Mixed isomers Purity Grade: Environmental Grade Formula Weight: 86.18 MDL Number: MFCD02179311 UN Number: UN1208
n-Hexane, for analysis, contains appr. 95% n-Hexane, ACROS Organics™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Purity Grade: For analysis Assay: ≥95.0% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
Hexane, Laboratory Reagent, ≥95%, Honeywell™
Vapor Pressure: 132mmHg (20°C), 256mmHg (37.7°C), 5.2psi (37.7°C) Vapor Density: 3 (vs air) Density: 0.659g/mL at 25°C Explosion Limit: 0.077 Flash Point: −26°C (−14.8°F) Ignition Point: 453°F Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 UN Number: UN1208 Assay: ≥95% Formula Weight: 86.18g/mol Purity Grade: Laboratory Reagent Chemical Name or Material: Hexane CAS: 110-54-3 Beilstein: 1730733 Refractive Index: n20/D 1.375(lit.) Melting Point: −95°C Boiling Point: 69°C Color: Colorless Physical State: Liquid
n-Hexane, For ACS analysis, MP Biomedicals™
Chemical Name or Material: n-Hexane Name Note: For ACS analysis CAS: 110-54-3 Physical Form: Liquid Assay Percent Range: 99.9% Molecular Formula: C6H14 Formula Weight: 86.18 Synonym: Hexane, Esani
Hexane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane), Honeywell
CAS Max %: <40.0000% Density: 0.659 - 0.673 g/cm3 Boiling Point: 68.7°C Solubility: 1.4% in Water at 20°C Formula Weight: 86.18 CAS Min %: >60.0000% Purity Grade: ACS Reagent Water: 0.01% max. Solubility Information: 1.4% in Water at 20°C CAS: 110-54-3 Synonym: N-HEXANE Flash Point: −26°C Assay Percent Notes: by GC Analysis n-Hexane Chemical Name or Material: Hexane CAS Max %: >60.00% Beilstein: 1730733 Name Note: ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.9% (>60% n-Hexane) Assay: >60% UN Number: UN1208
n-Hexane, >95% min., Ultra Scientific
Chemical Name or Material: n-Hexane CAS: 110-54-3 CAS Max %: 100.0 Assay Percent Range: >95% min.
Hexane, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99% (GC), Honeywell™ Riedel-de Haën™
Appearance: Liquid Color: 0 to 10 APHA Vapor Pressure: 5.2psi (37.7°C), 132mmHg (20°C), 256mmHg (37.7°C) Vapor Density: 3 (vs air) Density: 0.659g/mL at 25°C Explosion Limit: 0.077 Flash Point: −26°C (−14.8°F) Ignition Point: 453°F Residue after Evaporation: 0 to 0.001% Melting Point: −95°C Boiling Point: 69°C Refractive Index: n20/D 1.3745 to 1.3755 Titratable Acid: Free acid (as CH3COOH): ≤0.002%
Hexane, B&J Brand™, for GC, for pesticide residue analysis, non-spectrophotometric grade, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson
CAS Max %: <40.0000% Density: 0.659 - 0.673 g/cm3 Boiling Point: 68.7°C Formula Weight: 86.18 CAS Min %: >60.0000% Purity Grade: for pesticide residue analysis CAS: 110-54-3 Flash Point: −26°C Chemical Name or Material: Hexane CAS Max %: >60.00% Beilstein: 1730733 Name Note: B&J Brand™, for GC, for pesticide residue analysis, non-spectrophotometric grade, >99.9% (>60% n-Hexane) Packaging: Glass Bottle Assay: >99.9% UN Number: UN1208
Hexane, B&J Brand™, for HPLC, gradient grade, spectrophotometry (not for pesticide residue analysis), >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson
CAS Max %: <40.0000% Density: 0.659 - 0.673 g/cm3 Boiling Point: 68.7°C Solubility: 1.4% in Water at 20°C Formula Weight: 86.18 CAS Min %: >60.0000% Purity Grade: for HPLC, gradient grade Water: 0.01% max. Solubility Information: 1.4% in Water at 20°C CAS: 110-54-3 Synonym: N-HEXANE Flash Point: −26°C Chemical Name or Material: Hexane CAS Max %: >60.00% Beilstein: 1730733 Name Note: B&J Brand™, for HPLC, gradient grade, spectrophotometry (not for pesticide residue analysis), >99.9% (>60% n-Hexane) Packaging: Returnable Steel Drum Assay: >99.9% UN Number: UN1208
Hexane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson
CAS Max %: <40.0000% Density: 0.659 - 0.673 g/cm3 Boiling Point: 68.7°C Solubility: 1.4% in Water at 20°C Formula Weight: 86.18 CAS Min %: >60.0000% Purity Grade: ACS Reagent Water: 0.01% max. Solubility Information: 1.4% in Water at 20°C CAS: 110-54-3 Synonym: N-HEXANE Flash Point: −26°C Assay Percent Notes: by GC Analysis n-Hexane Chemical Name or Material: Hexane CAS Max %: >60.00% Beilstein: 1730733 Name Note: Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9% (>60% n-Hexane) Assay: >60% UN Number: UN1208
Heptane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (>99.0% n-Heptane), Honeywell Burdick & Jackson
Density: 0.684 g/cm3 Boiling Point: 98.4°C Formula Weight: 100.21 Purity Grade: for HPLC CAS: 142-82-5 Flash Point: −4°C Chemical Name or Material: Heptane CAS Max %: 100.0000% Beilstein: 1730763 Name Note: B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (>99.0% n-Heptane) Packaging: Glass Bottle Assay: >99.9% UN Number: UN1206
  spinner