Application Specific Hexanes
Hexanes (Pesticide), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexanes CAS: 110-54-3 Purity Grade: Pesticide Grade Purity Grade Notes: Also meets ACS specifications Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD02179311
Hexanes (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexanes CAS: 110-54-3 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥98.5% Assay Percent Range: Assay: 98.5% min by GC Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD00009520
Hexanes, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexanes Name Note: 98.5% CAS: 92112-69-1 CAS: 110-54-3 Purity Grade: For HPLC Assay: ≥98.5% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
Hexanes, mixed isomers, HPLC Grade, greater than99%, Alfa Aesar™
Chemical Name or Material: Hexanes CAS: 92112-69-1 CAS: 110-54-3 CAS: 96-14-0 CAS: 107-83-5 CAS: 96-37-7 CAS: 71-43-2 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD02179311
Hexanes (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Hexanes CAS: 110-54-3 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥98.5% Assay Percent Range: ≥98.5 % Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD02179311 Synonym: Naphtha Solvent
Hexanes, for residue analysis, ACROS Organics™
Chemical Name or Material: Hexanes CAS: 92112-69-1 CAS: 110-54-3 Purity Grade: For residue analysis Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
Hexanes (GC Resolv™), Fisher Chemical
Chemical Name or Material: Hexanes CAS: 110-54-3 Purity Grade: GC Resolv Purity Grade Notes: High purity solvent for sample prep prior to trace analysis by GC and/or GC/MS Assay Percent Range: 99.9% min by GC (aliphatic C6 hydrocarbons); 65% min ntohexane Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD00009520
Hexanes, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexanes CAS: 110-54-3 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay Percent Range: 99.9% min by GC Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD02179311
Hexanes UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Hexanes Name Note: (95% n-Hexane) CAS: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18
Hexane, CHROMASOLV™, ≥97%, LC-MS, Honeywell Riedel-de Haën™
CAS: 110-54-3 Chemical Name or Material: Hexane Density: 0.659g/mL at 25°C Assay: ≥97% Formula Weight: 86.18 Explosion Limit: 0.077 Flash Point: −26°C (−14.8°F) Health Hazard 2: P201-P210-P273-P301 + P310-P308 + P313-P331 Ignition Point: 453°F Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 RTECS Number: 203-777-6 UN Number: UN1208 Vapor Density: 3 (vs air) Vapor Pressure: 5.2 psi (37.7°C), 132 mmHg (20°C), 256 mmHg (37.7°C)
OmniSolv™ Hexanes, 98.5%, EMD Millipore
Chemical Name or Material: Hexanes CAS: 110-54-3 Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18
Hexanes, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexanes CAS: 110-54-3 Purity Grade: HPLC Grade Physical Form: Liquid Linear Formula: CH3(CH2)4CH3 with 2- and 3-methylpentanes and methylcyclopentane Formula Weight: 86.18
Hexanes, 98.5%, For GC Analysis, Non - UV, OmniSolv™, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexanes CAS: 110-54-3 Assay Percent Range: 98.50% Molecular Formula: C6 - H14 Synonym: Hexane
n-Hexane 95%, HPLC, EMD Millipore
Chemical Name or Material: n-Hexanes CAS: 110-54-3 Purity Grade: HPLC Grade Molecular Formula: C6H14 Formula Weight: 86.18 UN Number: 1208 Synonym: Hexane
OmniSolve Hexane, 99.8%, HR-GC Grade, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexane, 99.8% Name Note: HR-GC, OmniSolv™ CAS: 110-54-3 Purity Grade: HR-GC Physical Form: Liquid Assay Percent Range: 99.80% Linear Formula: CH3(CH2)4CH3 with 2- and 3-methylpentanes and methylcyclopentane Formula Weight: 86.18
Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Honeywell Riedel-de Haën™
Beilstein: 1730733 CAS: 110-54-3 Chemical Name or Material: Hexane Density: 0.659g/mL at 25°C Assay: ≥97.0% (GC) Boiling Point: 69°C Formula Weight: 86.18g/mol Melting Point: −95°C Purity Grade: CHROMASOLV™ EINECS Number: 203-777-6 Health Hazard 2: P201-P210-P273-P301 + P310-P308 + P313-P331 Health Hazard 3: H225-H304-H315-H336-H361f-H373-H411 Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 Molecular Formula: C6H14 Name Note: for HPLC Refractive Index: n20/D 1.375 RTECS Number: MN9275000 Synonym: n-Hexane Vapor Density: ˜3 (vs. air) Vapor Pressure: 256mmHg (37.7°C)
  spinner