Application Specific n- Hexane
n-Hexane, Alfa Aesar™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Physical Form: Liquid, Distilled In Glass Assay Percent Range: 95% min Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520
n-Hexane, 95%, for residue analysis, for trace analysis of polyaromatic hydrocarbons, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane CAS: 110-54-3 CAS Min %: 94.0 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥95% Assay Percent Range: 95% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, 95%, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane CAS: 110-54-3 (n-Hexane, 95%) CAS: 107-83-5 (2-Methylpentane, 2.3%) CAS: 96-14-0 (3-Methylpentane, 1.0%) Purity Grade: OPTIMA™ Purity Grade Notes: Also meets ACS specifications Assay Percent Range: 95% Molecular Formula: C6H14 Formula Weight: 86.17 MDL Number: MFCD00009520
n-Hexane, Environmental Grade, >95%, Alfa Aesar™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >95% Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520
n-Hexane, 95%, for residue analysis, ECD tested halocarbons free grade, ACROS Organics™
Chemical Name or Material: n-Hexane Name Note: ECD Tested CAS: 110-54-3 CAS Min %: 94.0 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥94% Assay Percent Range: 95% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, Spectrophotometric Grade, >95%, Alfa Aesar™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >95% Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520
Hexanes, for HPLC, contains ca. 95% n-Hexane, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexanes Name Note: (95% n-Hexane) CAS: 92112-69-1 CAS: 110-54-3 Purity Grade: For HPLC Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, 97%, for electronic use, residue free, ACROS Organics™
Chemical Name or Material: n-Hexane Name Note: Residue Free CAS: 110-54-3 CAS Min %: 96.0 CAS Max %: 100.0 Purity Grade: For electronic use Assay: ≥96% Assay Percent Range: 97% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-hexane, 95+%, for HPLC, ACROS Organics™
Chemical Name or Material: n-Hexane Name Note: For HPLC CAS: 110-54-3 CAS Min %: 95.0 CAS Max %: 100.0 Purity Grade: For HPLC Assay: ≥95.0% Assay Percent Range: 95+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Synonym: Hex
n-Hexane, for residue and pesticides anal., ca. 95% n-Hexane, ACROS Organics™
Chemical Name or Material: n-Hexane Name Note: ECD Tested, for Pesticide Analysis, Residue <0.0003% CAS: 110-54-3 Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane Name Note: OmniPure™ CAS: 110-54-3 Assay Percent Range: 95% Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18
n-Hexane For gas chromatography ECD and FID SupraSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexane CAS: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 UN Number: 1208
n-Hexane For gas chromatography MS SupraSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexane CAS: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 UN Number: 1208
n-Hexane For spectroscopy Uvasol™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexane CAS: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 UN Number: 1208
Hexane, CHROMASOLV™, Honeywell Riedel-de Haën™
Beilstein: 1730733 CAS: 110-54-3 Chemical Name or Material: Hexane Density: 0.659g/mL at 25°C Formula Weight: 86.18g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.077 Flash Point: −26°C (−14.8°F) Ignition Point: 453°F Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 UN Number: UN1208 Vapor Density: 3 (vs air) Vapor Pressure: 5.2 psi (37.7°C), 256 mmHg (37.7°C), 132 mmHg (20°C)
Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Honeywell Riedel-de Haën™
Beilstein: 1730733 CAS: 110-54-3 Chemical Name or Material: Hexane Density: 0.659g/mL at 25°C Assay: ≥97.0% (GC) Boiling Point: 69°C Formula Weight: 86.18g/mol Melting Point: −95°C Purity Grade: CHROMASOLV™ Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 Molecular Formula: C6H14 Name Note: for HPLC Refractive Index: n20/D 1.375 Synonym: n-Hexane Vapor Density: ˜3 (vs. air) Vapor Pressure: 256mmHg (37.7°C)
  spinner