Formamide
Formamide (Certified ACS), Fisher Chemical
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.041 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N Synonym: Methanamide PubChem CID: 713 ChEBI: CHEBI:48431
Formamide, 99%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide Name Note: DNase, RNase and Protease Free CAS: 75-12-7 Purity Grade: For molecular biology Assay: 99% Assay Percent Range: 99% Molecular Formula: CH3NO Linear Formula: HCONH2 MDL Number: MFCD00007941
Formamide, 99.5+%, extra pure, ACROS Organics™
Chemical Name or Material: Formamide Name Note: 99.5+% CAS: 75-12-7 Purity Grade: Extra Pure Assay: ≥99.5% Assay Percent Range: 99.5+% Molecular Formula: CH3NO Linear Formula: HCONH2 MDL Number: MFCD00007941
Formamide, 99+%, for spectroscopy, ACROS Organics™
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: For spectroscopy Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: CH3NO Linear Formula: HCONH2 MDL Number: MFCD00007941
Formamide (Molecular Biology), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Molecular Biology Grade Physical Form: Liquid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: CH3NO Linear Formula: H2NCH:O
Formamide, 99.5%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: For analysis Assay: 99.5% Assay Percent Range: 99.5% Molecular Formula: CH3NO Linear Formula: HCONH2 MDL Number: MFCD00007941
Formamide (≥99.5%), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.041 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N PubChem CID: 713 ChEBI: CHEBI:48431
Formamide, 99.6%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: CH3NO Linear Formula: HCONH2 MDL Number: MFCD00007941
Formamide OR, J.T.Baker™
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.041 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N PubChem CID: 713 ChEBI: CHEBI:48431
Formamide, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide Name Note: For ACS analysis CAS: 75-12-7 Purity Grade: ACS/EP/Reagent Grade Physical Form: Slightly Viscous Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3NO Linear Formula: HCONH2 Synonym: Formic acid amide, Methane amide
Formamide, Polarographic and Nonaqueous, Titration Grade, ACROS Organics™
CAS: 75-12-7 Molecular Formula: CH3NO Molecular Weight (g/mol): 45.041 MDL Number: MFCD00007941 InChI Key: ZHNUHDYFZUAESO-UHFFFAOYSA-N PubChem CID: 713 ChEBI: CHEBI:48431
  spinner