Multicompendial 1- Propanol
n-Propyl Alcohol, WORLD/GMP, ACS/FCC/EP Grade, PHARMCO-AAPER
Chemical Name or Material: n- Propyl Alcohol CAS: 71-23-8 Physical Form: Liquid Molecular Formula: C3H8O Formula Weight: 60.1 Synonym: 1-Propanol, Propan-1-ol
  spinner