Multicompendial Ethyl Acetate
Ethyl Acetate (NF), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: BP/EP/FCC/JP/NF/USP Assay: 99.0 to 100.5% (NF) Assay Percent Range: 99.0 to 100.5 % Molecular Formula: C4H8O2 Linear Formula: CH3COOC2H5 MDL Number: MFCD00009171 Synonym: Acetic Acid Ethyl Ester
Ethyl Acetate, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, ≥99.5%, Honeywell™
Density: 0.8945 g/cm3 Boiling Point: 77°C Solubility: 870% in Water at 20°C Formula Weight: 88.11 Purity Grade: for HPLC Water: 0.03% max. Solubility Information: 870% in Water at 20°C CAS: 141-78-6 Synonym: EtOAc MDL Number: MFCD00009171 Flash Point: −4°C Chemical Name or Material: Ethyl Acetate CAS Max %: 100.0000% Beilstein: 506104 Name Note: ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, ≥99.5% Assay: >99.5% UN Number: UN1173
Ethyl Acetate, puriss., ≥99.5% (GC); Honeywell™ Riedel-de Haën™
Vapor Pressure: 73mmHg (20°C) Vapor Density: 3 (20°C, vs air) Density: 0.898 to 0.902g/mL at 20°C, 0.894 to 0.898g/mL at 25°C Explosion Limit: 2.2 to 11.5% (38°F) Flash Point: −3°C (26.6°F) Ignition Point: 801°F Linear Formula: CH3COOC2H5 Name Note: Meets analytical specification of Ph. Eur.BP, NF MDL Number: MFCD00009171 UN Number: UN1173 Assay: ≥99.5% (GC) Formula Weight: 88.11g/mol Purity Grade: puriss. Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Beilstein: 506104 Color: Colorless Physical State: Liquid Boiling Point: 76.5° to 77.5°C Melting Point: −84°C
Ethyl Acetate, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Physical Form: Liquid Molecular Formula: C4H8O2 Formula Weight: 88.1 Synonym: Acetic Acid Ethyl Ester, Ethyl Ethanoate, Ethyl Acetic Ester
  spinner