Multicompendial Toluene
Toluene, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.8%, Honeywell™
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.141 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: antisal 1a, benzene, methyl, methacide, methylbenzene, methylbenzol, monomethyl benzene, phenylmethane, tolu-sol, toluen, toluol PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
  spinner