p- Xylene
p-Xylene (Certified), Fisher Chemical™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: 1,4-dimethylbenzene, 4-methyltoluene, 4-xylene, benzene, 1,4-dimethyl, chromar, p-dimethylbenzene, p-methyltoluene, p-xylene, p-xylol, para-xylene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C=C1)C
p-Xylene, 99+%, extra pure, ACROS Organics™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: 1,4-dimethylbenzene, 4-methyltoluene, 4-xylene, benzene, 1,4-dimethyl, chromar, p-dimethylbenzene, p-methyltoluene, p-xylene, p-xylol, para-xylene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C=C1)C
p-Xylene, 99%, pure, ACROS Organics™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 MDL Number: MFCD00008556 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: 1,4-dimethylbenzene, 4-methyltoluene, 4-xylene, benzene, 1,4-dimethyl, chromar, p-dimethylbenzene, p-methyltoluene, p-xylene, p-xylol, para-xylene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C=C1)C
p-Xylene, 99%, for HPLC, ACROS Organics™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: 1,4-dimethylbenzene, 4-methyltoluene, 4-xylene, benzene, 1,4-dimethyl, chromar, p-dimethylbenzene, p-methyltoluene, p-xylene, p-xylol, para-xylene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C=C1)C
p-Xylene, Reagent, 98% min., MilliporeSigma™
CAS: 106-42-3 Molecular Formula: C8H10 Molecular Weight (g/mol): 106.168 InChI Key: URLKBWYHVLBVBO-UHFFFAOYSA-N Synonym: 1,4-dimethylbenzene, 4-methyltoluene, 4-xylene, benzene, 1,4-dimethyl, chromar, p-dimethylbenzene, p-methyltoluene, p-xylene, p-xylol, para-xylene PubChem CID: 7809 ChEBI: CHEBI:27417 IUPAC Name: 1,4-xylene SMILES: CC1=CC=C(C=C1)C
  spinner