Lupus Testing
ASI™ SLE Test Kit
Detects anti-DNP antibodies associated with Systemic Lupus Erythematosus. The ASI SLE Slide Test is a latex slide agglutination assay for qualitative and semiquantitative detection of anti-deoxyribonucleoprotein (anti-DNP) in human serum.
Alere ANA HEp-2 Test System
Pre-standardized assay designed for qualitative and semi-quantitative detection of anti-nuclear antibodies
Bio/Data™ Lupus Anticoagulant Confirmation Reagent (LA-CR)
Designed to Perform platelet neutralization procedure (PNP)
Stanbio™ RaPET™ SLE Test Kit
Rapid Particle Enhanced Technology for superior macroagglutination
  spinner