Clinical Chemistry Standards
Abnova™ Exosome Standard (SK-N-SH cell line)
This product is purified and lyophilized exosomes obtained from SK-N-SH cell line (human neuroblastoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Ortho-Clinical Diagnostics™ VITROS™ 250/350 Calibrators
Calibrators for Ortho Clinical VITROS 250/350
Pointe Scientific General Chemistry Calibrators
Liquid-stable, serum-based calibrator sets contain both Total and Direct Bilirubin values
Stanbio™ Cholesterol Standard
For use in chemical/clincal testing
Thermo Scientific™ Clinical Chemistry Standards
Perform in vitro diagnosis with ammonia, HDL cholesterol and lithium clinical chemistry standards.
Abnova™ Exosome Standard (BPH-1 cell line)
This product is purified and lyophilized exosomes obtained from BPH-1 cell line (human prostatic hyperplasia) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (DAUD1 cell line)
This product is purified and lyophilized exosomes obtained from DAUD1 cell line (human burkitt lymphoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (A549 cell line)
This product is purified and lyophilized exosomes obtained from A549 cell line (human lung carcinoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Fluorescent Exosome Standard (HCT116 cell line) (Green dye)
This product is purified exosomes obtained from HCT116 cell line (human colon carcinoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Fluorescent Exosome Standard (DAUD1 cell line) (Green dye)
This product is purified exosomes obtained from DAUD1 cell line (human burkitt lymphoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Fluorescent Exosome Standard (U87 MG cell line) (Green dye)
This product is purified exosomes obtained from U87 MG cell line (human glioblastoma astrocytoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Exosome Standard (B16F10 cell line)
This product is purified and lyophilized exosomes obtained from B16F10 cell line (mouse melanoma cell line) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (PC3 cell line)
This product is purified and lyophilized exosomes obtained from PC3 cell line (human prostrate adenocarcinoma grade IV) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (K-562 cell line)
This product is purified and lyophilized exosomes obtained from K-562 cell line (human pleural effusion, leukemia chronic myelogenous) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (U87 MG cell line)
This product is purified and lyophilized exosomes obtained from U87 MG cell line (human glioblastoma astrocytoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (BLCL21 cell line)
This product is purified and lyophilized exosomes obtained from BLCL21 cell line (EBV transformed lymphoblastoid B cells) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (MM1 cell line)
This product is purified and lyophilized exosomes obtained from MM1 cell line (human melanoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Fluorescent Exosome Standard (COLO1 cell line) (Green dye)
This product is purified exosomes obtained from COLO1 cell line (human colon carcinoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Fluorescent Exosome Standard (A549 cell line) (Green dye)
This product is purified exosomes obtained from A549 cell line (human lung carcinoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Fluorescent Exosome Standard (SK-N-SH cell line) (Green dye)
This product is purified exosomes obtained from SK-N-SH cell line (human neuroblastoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Fluorescent Exosome Standard (BLCL21 cell line) (Green dye)
This product is purified exosomes obtained from BLCL21 cell line (EBV transformed lymphoblastoid B cells) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Fluorescent Exosome Standard (B16F10 cell line) (Green dye)
This product is purified exosomes obtained from B16F10 cell line (mouse melanoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Fluorescent Exosome Standard (BPH-1 cell line) (Green dye)
This product is purified exosomes obtained from BPH-1 cell line (human prostatic hyperplasia) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
Abnova™ Exosome Standard (HCT116 cell line)
This product is purified and lyophilized exosomes obtained from HCT116 cell line (human colon carcinoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Exosome Standard (COLO1 cell line)
This product is purified and lyophilized exosomes obtained from COLO1 cell line (human Colon Carcinoma) and can be used as control standards for multiple applications including FACS, WB, ELISA and as calibration standards for quantitation of exosome-derived markers from biological samples.
Abnova™ Fluorescent Exosome Standard (MM1 cell line) (Green dye)
This product is purified exosomes obtained from MM1 cell line ((human melanoma) and labelled with fluorescent green dye. It can be used for extracellular vesicle tracking in fluorescence microscopy experiments.
  spinner