DOA Controls
Alere iScreen™ Urine Drug Control Kit
Use these external controls to validate test performance and expected results. Positive and negative controls for use with Alere urine drug screens.
MEDTOX™ Negative Control
Designed to monitor the performance of MEDTOX™ drugs of abuse test devices. MEDTOX™ Negative Controls are compatible with all quantitative and qualitative drug detection procedures, which are sufficiently sensitive to detect the control constituents.
MEDTOX™ Positive Control for Profile-V MEDTOXScan™ test systems
Monitors the performance of Profile-V MEDTOXScan™ drugs of abuse test devices. MEDTOX™ Positive Controls for Profile-V MEDTOXScan test systems are compatible with all quantitative and qualitative drug detection procedures that are sufficiently sensitive to detect the control constituents.
Sciteck™ SVT™ Adulteration Specific Gravity Reagent
For in vitro diagnostic use to determine specific gravity of solution
Alere iScreen™ Dx Dip Card, Multi Drug Configuration
For rapid urine drug screening (multiple)
Sciteck™ SVT™ Adulteration pH Reagent
For in vitro diagnostic use to determine pH of a solution
Sciteck™ SVT™ Adulteration SAM-Set™ Calibrator Series
For use in validity tests for pH, creatinine, nitrite, and refractometer (specific gravity)
MEDTOX™ MEDTOXScan™ Cleaning Cassette
Use this Cleaning Cassette in conjunction with MEDTOXScan QC Test Devices to clean the internal components of the MEDTOXScan reader. The MEDTOXScan™ Cleaning Cassette is solely intended to clean the color Contact Imaging Sensor (CIS) in the MEDTOXScan™ Reader.
Sciteck™ SVT™ Adulteration Creatinine Calibrators
For use with automated systems for drugs of abuse adulteration testing
Alere iScreen™ External Drug Controls
Used as quality control urine to monitor precision of laboratory urine toxicology testing procedures for different analytes
MEDTOX™ MEDTOXScan™ Positive and Negative QC Test Device
Detect errors associated with the MEDTOXScan™ Reader and a contaminated contact imaging sensor (CIS). MEDTOXScan™ Positive and Negative QC Test Devices verify that the CIS cleaning procedure using the MEDTOXScan™ Cleaning Cassette effectively removed any contamination.
MEDTOX™ Positive Control
Designed to monitor the performance of MEDTOX™ drugs of abuse test devices. MEDTOX™ Positive Controls are compatible with all quantitative and qualitative drug detection procedures, which are sufficiently sensitive to detect the control constituents.
Alere Integrated E-Z Split Key™ Cup A.D. with Built-in Validity Drug Screening
Cup with key activated plunger allows to run device as per convenience
Alere QED™ A150 Saliva Alcohol Test Device
Simple-to-use alcohol test that runs like a thermometer and gives quantitative results in few minutes
Alere iScreen™ OFD Cotinine Test Device
For rapid determination of smoking status with oral fluid
Alere iScreen™ Saliva Alcohol Test Strips
Convenient and rapid preliminary test that detects saliva alcohol concentration (SAC)
Alere AlcoMate™ Premium 7000 Breath Alcohol Tester Kit
DOT approved tester with one button operation that requires no calibration service to use
Alere iScreen™ Dx Dip Cards, Single Drug Configuration
For rapid urine drug screening (single)
Alere iScreen™ Adulteration Test Strips
Used for testing six different parameters
Sciteck™ SVT™ Adulteration Nitrite Calibrators
For laboratory use only to determine nitrite content of a solution
Alere iCassette™ Single Test Drug Screen Cassettes
Rapid and accurate screening of drugs in minutes
Sciteck™ SVT™ Adulteration Halogen (Bleach) Reagent
For laboratory use only to determine halogen (e.g. bleach, etc.) content of a solution
Sciteck™ SVT™ Adulteration Chromate Reagent
For use with automated systems for drugs of abuse adulteration testing
Sciteck™ UR-N-Control™ Tri-Level Adulteration QC Set
For use with automated analyzer systems performing adulteration detection
Sciteck™ SVT™ Adulteration Aldehydes Reagent
For use with automated systems for drugs of abuse adulteration testing
Sciteck™ SVT™ Adulteration Quality Control Series
For use in validity tests for pH, creatinine, nitrite, and oxidants
Sciteck™ SVT™ Adulteration Peroxidase Reagent
For in vitro diagnostic use to determine peroxidase in urine
Sciteck™ SVT™ Adulteration Nitrite Reagents
For laboratory use only to determine nitrite content of a solution
  spinner