Albumin Testing
Roche Diagnostics Chemstrip™ Micral™ Albumin Test
Two easy steps — results in one minute
Siemens Healthineers Clinitek™ Microalbumin 2 Reagent Strips Available on GSA/VA Contract for Federal Government customers only.
For use with Clinitek 50, 100, Status Family of Urine Chemistry Analyzers
Stanbio™ LiquiColor™ Albumin Test
For Bromocresol Green reaction
Germaine™ Laboratories MicroAlbumin 2-1 Combo Strips
Germaine™ Laboratories MicroAlbumin 2-1 Combo Dipstick reagent strips are for the qualitative detection of Microalbumin and Creatinine
Ricca Chemical Exton Qualitative Reagent
For the Qualitative Detection of Albumin in Urine.
Abnova™ BCG Albumin Assay Kit Available on GSA/VA Contract for Federal Government customers only.
BCG Albumin Assay Kit is a quantitative colorimetric albumin determination at 620 nm.
Abnova™ BCP Albumin Assay Kit Available on GSA/VA Contract for Federal Government customers only.
BCP Albumin Assay Kit is a quantitative colorimetric albumin determination at 610 nm.
Germaine™ Laboratories AimStrip MACRE Strips
Germaine™ Laboratories AimStrip™ MACRE Strips are dipstick reagent strips for the qualitative detection of Microalbumin and Creatinine
  spinner