Antiphospholipid Syndrome Testing Kits
Phadia™ β 2-Glycoprotein I Antibodies for Antiphospholipid Syndrome
Intended for the in vitro quantitative measurement of antibodies directed to β2-Glycoprotein I in human serum and plasma to aid in the diagnosis of antiphospholipid syndrome (APS) and to evaluate the thrombotic risk in patients with systemic lupus erythematosus (SLE).
  spinner