Cardiac Testing
Tosoh Bioscience AIA-PACK™ Cardiac Markers Assays: Assay Test Cups
Pre-measured, single unit-dosed dry reagents
Quidel Triage MeterPro BNP Panel 25/PK
Triage BNP is a rapid, point-of-care CLIA-Waived fluorescence immunoassay used with the Triage MeterPro. It is used to measure B-type natriuretic peptide (BNP) in EDTA anticoagulated whole blood or plasma specimens.
Quidel Triage MeterPro Cardiac Panel 25/PK
Triage Cardiac Panel is a rapid, point of care fluorescence immunoassay for use on the Triage MeterPro. This test is designed to determine the level of creatine kinase MB (CK-MB), myoglobin and troponin I in EDTA anticoagulated whole blood or plasma specimens.
Quidel Triage MeterPro Cardiac Panel Troponin/CK-MB 25/PK
Triage Cardiac Panel is a rapid, point of care fluorescence immunoassay for use on the Triage MeterPro. This test is designed to determine the level of creatine kinase MB (CK-MB) and troponin I in EDTA anticoagulated whole blood or plasma specimens.
Quidel Triage MeterPro Cardiac Panel Troponin 25/PK
Triage Cardiac Panel is a rapid, point of care fluorescence immunoassay for use on the Triage MeterPro. This test is designed to determine the level of troponin I in EDTA anticoagulated whole blood or plasma specimens. This can be used as an aid in the diagnosis of myocardial infarction (injury).
Tosoh Bioscience AIA-PACK™ Cardiac Markers Assays: Calibrators
Pre-measured, single unit-dosed dry reagents
Quidel Triage MeterPro Combo Cardiac Panel/BNP 50 total tests
See Cardiac Panel and BNP Test for more detail
PTS Diagnostics CardioChek™ PA Analyzer
Cost-efficient, user-friendly diagnostic point-of-care analyzer, which can accurately determine multiple patient values
Tosoh Bioscience AIA-PACK™ Cardiac Markers Assays: Solutions
Pre-measured, single unit-dosed dry reagents
PTS Diagnostics Accessories for CardioChek™ PA Analyzer
Experience increased versatility and usefulness with these accessories
LifeSign Lifesign MI™ Cardiac Markers Test Kit
Lifesign MI™ tests are for rapid detection of cardiac markers
PTS Diagnostics CardioChek™ ChekMate™ Quality Control kit
A two level, dry strip quality control system designed to verify the performance of the CardioChek PA device
PTS Diagnostics CardioChek™ Health Risk Assessment (HRA)
Stay Connected to Your Health with the CardioChek Health Risk Assessment (HRA)
Quidel Triage MeterPro Combo Cardiac Panel, BNP, D-DIMER 75 total tests
See Cardiac Panel, BNP Test and D-Dimer Test for more detail
Quidel Triage MeterPro Combo Troponin, Troponin/CK-MB with BNP, and D-Dimer 75 total tests
See Cardiac Panel (97022HS), BNP Test and D-Dimer Test for more detail
PTS Diagnostics CardioChek™ Health Risk Assessment (HRA)
Stay Connected to your health with the CardioChek Health Risk Assessment (HRA)
Quidel Triage MeterPro Combo Troponin/CK-MB and BNP 50 total tests
See Cardiac Panel (97022HS) and BNP Test for more detail
Quidel Triage MeterPro Combo Troponin, BNP, D-Dimer 75 total tests
See Cardiac Panel (97021HS), BNP Test and D-Dimer Test for more detail
Quidel Triage MeterPro Cardiac Troponin only plus BNP 25/PK
See Cardiac Panel (97021HS), and BNP Test for more detail
Vernier Vernier EKG Electrodes Products for Science Education
Replacement package of 100 electrodes for use with EKG sensor.
  spinner