HIV Testing
Alere Determine™ HIV-1/2 Ag/Ab Combo Test Available on GSA/VA Contract for Federal Government customers only.
Diagnose HIV infection earlier — get medical care sooner. Rapid point-of-care test simultaneously detects both HIC-1/2 antibodies and HIV-1 p24 antigen.
Trinity Biotech Uni-Gold™ Recombignen™ HIV-1/2 Available on GSA/VA Contract for Federal Government customers only.
Suitable for use in appropriate multi-test algorithms designed for the statistical validation of rapid HIV test results
Alere Determine™ HIV-1/2 Ag/Ab Combo Blood Test Available on GSA/VA Contract for Federal Government customers only.
First 4th generation rapid point-of-care test that detects both HIV-1/2 antibodies and free HIV-1 p24 antigen. Enables health care providers to diagnose HIV infection earlier allowing individuals to seek medical care sooner.
Chembio HIV 1/2 STAT-PAK™ Assay
Easy-to-perform, single-use diagnostic tests for the rapid (15 minutes), visual detection of antibodies to HIV 1 and HIV 2 at point-of-care (POC)
Chembio SURE CHECK™ HIV 1/2 Assay
Unique, easy-to-use, self-contained collection and testing device for the rapid, visual detection of antibodies to HIV-1 and HIV-2 in whole blood, serum or plasma.
Alere Determine™ External Controls for HIV-1/2 Ag/Ab Combo Test Available on GSA/VA Contract for Federal Government customers only.
Diagnose HIV infection earlier — get medical care sooner. Rapid point-of-care test simultaneously detects both HIC-1/2 antibodies and HIV-1 p24 antigen.
Chembio DPP™ HIV 1/2 Assay
Patented DPP™ technology for the rapid detection of HIV 1 and HIV 2 antibodies in oral fluid and all blood matrices
Chembio HIV Rapid Test Control Pack
To be used in conjunction with HIV 1/2 STAT-PAK™ (part number 60-9505-1) and SURE CHECK™ HIV 1/2 (part number 60-9507-0) Assays
Chembio DPP™ HIV 1/2 Rapid Test Control Pack
Control Pack for Chembio DPP™ HIV 1/2 Assay
Zeptometrix NATtrol™ HIV Type 1 Strain: IIIB Linearity Panel Available on GSA/VA Contract for Federal Government customers only.
Designed to evaluate the performance of nucleic acid tests for determination of the presence of HIV-1 RNA
Alere Clearview™ HIV 1/2 Control Kit
For use with Alere Clearview COMPLETE HIV 1/2 and Alere Clearview HIV 1/2 STAT-PAK™ test kits
Alere Clearview™ HIV 1/2 Rapid Test Kits
Single-use tests for detecting HIV-1 and HIV-2 antibodies in human whole blood, serum or plasma samples
Alere Clearview™ HIV 1/2 STAT-PAK™
A rapid, qualitative screening test for the detection of antibodies to HIV-1 and HIV-2 in human whole blood, serum and plasma
  spinner