FDP Assay Reagents
Pacific Hemostasis™ FDP Assay Available on GSA/VA Contract for Federal Government customers only.
Fisherbrand™ Pacific Hemostasis FDP Assay is a fast, easy-to-use latex agglutination assay for use in measuring fibrinogen degradation products (FDP) in serum and urine.
  spinner