Fibrinogen Testing
Thermo Scientific™ Pacific Hemostasis™ Imidazole Buffered Saline
Fisherbrand™ Pacific Hemostasis Fibrinogen/Fibrin Analysis is a complete line of assays used to screen and quantify plasma fibrinogen based on the Clauss method.
  spinner