aPPT Reagents Activated Partial Thromboplastin Time
Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents Available on GSA/VA Contract for Federal Government customers only.
Identify acquired or congenital disorders to the intrinsic and common pathways.
Fisher Diagnostics™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (aPTT) Reagents Available on GSA/VA Contract for Federal Government customers only.
aPTT reagents provide sensitivity to intrinsic and common pathway factors, heparin and lupus anticoagulant
  spinner