Flu and Virus Testing
Fisher Healthcare™ Sure-Vue™ Mono Latex Test Kit Available on GSA/VA Contract for Federal Government customers only.
Qualitative and semi-quantitative results
Fisher Healthcare™ Sure-Vue™ Rubella Latex Test Kit Available on GSA/VA Contract for Federal Government customers only.
Requires a dilution step for 10 IU sensitivity
Fisher Healthcare™ Sure-Vue™ SELECT Rubella Latex Test Available on GSA/VA Contract for Federal Government customers only.
Does not require a dilution step for 10 IU sensitivity
Fisher HealthCare™ Sure-Vue™ Signature Mono Test Kit Available on GSA/VA Contract for Federal Government customers only.
Rapid, lateral flow test, waived for whole blood.
Thermo Scientific™ Xpect™ Rotavirus Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Rotavirus from human fecal samples.
Thermo Scientific™ Remel™ Color Slide™ II Mononucleosis Test Available on GSA/VA Contract for Federal Government customers only.
Rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
Thermo Scientific™ Xpect™ Flu A & B Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens, and get results in 15 minutes.
Thermo Scientific™ Remel™ Xpect™ Rotavirus Control Kit Available on GSA/VA Contract for Federal Government customers only.
Remel Xpect and Rotavirus Control Kit is used with Xpect Rotavirus.
Abbott™ ID NOW RSV Test Kit
Highly sensitive rapid diagnostic tests for RSV are needed to help guide management in the clinical setting. Combines the benefits of speed and accuracy on an easy to use system.
Abbott™ ID NOW Influenza A and B 2
The fastest molecular flu test just got faster and even better. Now with early detection and positive results in as few as 5 minutes, allowing you to make effective clinical decisions sooner. Abbott ID NOW is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.
Thermo Scientific™ Xpect™ RSV Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Respiratory Syncytial Virus (RSV) from nasopharyngeal specimens in neonatal and pediatric patients and get results in 15 minutes.
Fisher Healthcare™ Sure-Vue™ RSV Test Kit Available on GSA/VA Contract for Federal Government customers only.
Alternative procedure for bloody and mucoid samples
Alere BinaxNOW™ RSV Lateral Flow Test Kits Available on GSA/VA Contract for Federal Government customers only.
Rapid, one-step lateral flow test for Respiratory Syncytial Virus (RSV)
Thermo Scientific™ BinaxNOW™ RSV CLIA Waived Test Available on GSA/VA Contract for Federal Government customers only.
Detect RSV in 15 minutes with BinaxNOW™ CLIA Waived RSV, a rapid, one-step lateral flow test.
Quidel™ Sofia™ RSV FIA Immunofluorescence Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigens
Quidel Sofia™ Influenza A+B FIA Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid differential diagnosis of acute influenza A and influenza B viral infections
BD Veritor™ Plus Analyzer
Point of Care detection of infectious diseases such as influenza A and B, respiratory syncytial virus (RSV) and group A strep. BD Veritor Plus Analyzer provides lab-quality immunoassay test results within minutes.
BD Veritor™ System for the Rapid Detection of Flu A+B
Unique adaptive-read technology helps reduce false-positive results
BD Veritor System RSV (Respiratory Syncytial Virus) Test Kit
Provides objective results on an easy to read digital display, unlike current rapid RSV tests, which requires subjective visual test interpretation to determine positive or negative test result
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel, Respiratory Virus Available on GSA/VA Contract for Federal Government customers only.
An immunofluorescent test used for 7 respiratory viuses: influenza A and B; adenovirus; parainfluenza 1, 2, 3; and RSV in shell vial pre-CPE assays and tube cultures.
Sekisui Diagnostics OSOM™ Mono Test Kit Available on GSA/VA Contract for Federal Government customers only.
Simple, in-office test for the detection of mononucleosis
Thermo Scientific™ Remel™ Xpect™ RSV Control Kit Available on GSA/VA Contract for Federal Government customers only.
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
Quidel QuickVue™ RSV Test Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid, qualitative detection of RSV antigen directly from nasopharyngeal swab or aspirate specimens from symptomatic pediatric patients
Sekisui Diagnostics OSOM™ Ultra Flu A and B Rapid Test
OSOM Ultra Flu A&B is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens
Meridian Bioscience™ ImmunoCard™ STAT! Rotavirus Test Kit Available on GSA/VA Contract for Federal Government customers only.
No centrifugation required; simply dilute specimen and add to test device
Quidel QuickVue™ Influenza A+B Test Available on GSA/VA Contract for Federal Government customers only.
Aids in the rapid differential diagnosis of acute influenza A and B virus infection
  spinner