Influenza Testing
Alere i Influenza A and B Available on GSA/VA Contract for Federal Government customers only.
Provides molecular results in less than 15 minutes, allowing you to make effective clinical decisions sooner. Alere™ i is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.
Abbott™ Alere™ i Influenza A and B 2
The fastest molecular flu test just got faster and even better. Now with early detection and positive results in as few as 5 minutes, allowing you to make effective clinical decisions sooner. Alere™ i is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.
Thermo Scientific™ Xpect™ Flu A & B Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens, and get results in 15 minutes.
BD Veritor™ Plus Analyzer
Point of Care detection of infectious diseases such as influenza A and B, respiratory syncytial virus (RSV) and group A strep. BD Veritor Plus Analyzer provides lab-quality immunoassay test results within minutes.
Quidel™ Sofia™ Influenza A+B FIA Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid differential diagnosis of acute influenza A and influenza B viral infections
BD Veritor™ System for the Rapid Detection of Flu A+B
Unique adaptive-read technology helps reduce false-positive results
Quidel™ QuickVue™ Influenza A+B Test Available on GSA/VA Contract for Federal Government customers only.
Aids in the rapid differential diagnosis of acute influenza A and B virus infection
Sekisui Diagnostics OSOM™ Ultra Flu A and B Rapid Test
OSOM Ultra Flu A&B is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens
BD Veritor™ Plus Analyzer with Flu A+B CLIA Waived Flu Assay Kits (2 x 30)
Point of Care lab-quality detection of influenza A and B. BD Veritor™ Plus Analyzer with Flu A+B CLIA Waived Flu Assay Kits (2 x 30) includes an advanced chromatographic instrument and two immunoassay kits to detect antigens from respiratory specimens of symptomatic patients in 10 minutes.
Quidel™ Solana™ Flu Assay Startup Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid differential diagnosis of acute influenza A and influenza B viral infections.
Meridian Bioscience™ ImmunoCard STAT!™ FLU A&B
Meridian Bioscience ImmunoCard™ STAT! FLU A&B is a rapid Flu test for Nasal and NP swabs
Zeptometrix NATtrol™ Influenza/RSV Negative Control
Designed for use in evaluating and monitoring the performance of nucleic acid tests for determination of the presence of respiratory virus nucleic acids
Zeptometrix NATtrol™ Influenza External Run Controls Available on GSA/VA Contract for Federal Government customers only.
Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable
LifeSign Status Flu A and B Qualitative Test Kit
LifeSign Status Flu A and B Qualitative Test Kit is an in vitro rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.
Zeptometrix NATtrol™ Influenza A/B Positive Control
Designed for use in evaluating and monitoring the performance of nucleic acid tests for determination of the presence of respiratory virus nucleic acids
Sekisui Diagnostics Silaris™ Influenza A&B Starter Pack
Start Smart with the CLIA-Waived Silaris Influenza Starter Pack includes 1 Silaris Dock and 2 Silaris Influenza A&B Test.
Quidel™ Sofia™ 2 Flu A+B Starter Kit
Small bench top analyzer that uses advanced fluorescence detection with an ultraviolet LED energy source. Sofia 2 Fluorescent Immunoassay Analyzer collects hundreds of data points on the test strip contained inside of a Sofia FIA Test Cassette.
Thermo Scientific™ Remel™ Xpect™ Flu A and B Control Swab Available on GSA/VA Contract for Federal Government customers only.
Remel Xpect and Flu A & B Control Swabs are used as external quality control material for the Xpect Flu A and B test.
Alere BinaxNOW™ Flu A/B Swabs Available on GSA/VA Contract for Federal Government customers only.
Accurate differentiation between influenza A and influenza B using one step on one test
Germaine™ Laboratories, Inc. BioSign™ Flu A/B Kit
An In vitro rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.
BD CLIA Waived (Physician) Combo Reader Pack
Includes two packs of B256045 (Flu A + B Test Kit) and one pack of B256055 (Veritor System Reader)
BD Veritor™ Plus Analyzer with Flu A+B CLIA Moderate Complex Flu Assay Kits (2 x 30)
Point of Care lab-quality detection of influenza A and B. BD Veritor™ Plus Analyzer with Flu A+B CLIA Moderate Complex Flu Assay Kits (2 x 30) includes an advanced chromatographic instrument and two immunoassay kits to detect antigens from respiratory specimens of symptomatic patients in 10 min.
Quidel™ Sofia™ Starter Kit
Includes Sofia Barcode scanner with user manual and installation guide and one Influenza A+B FIA kit
Sekisui Diagnostics Silaris™ Influenza A&B Control  Kit
The Silaris™ Influenza  A&B Control  Kit is intended for in vitro diagnostic use in quality control testing with the Silaris Influenza A&B Test.
Quidel™ QuickVue™ Influenza Test Kit
Rapid qualitative detection of influenza type A and B antigens
Thermo Scientific™ Oxoid™ IMAGEN™ Parainfluenza Group Available on GSA/VA Contract for Federal Government customers only.
Oxoid IMAGEN Parainfluenza Group detects and confirms parainfluenza virus antigens in nasopharyngeal aspirates and cell culture preparations.
Thermo Scientific™ Oxoid™ IMAGEN™ Parainfluenza Typing Available on GSA/VA Contract for Federal Government customers only.
Oxoid IMAGEN Parainfluenza Typing detects and differentiates Parainfluenza virus Types 1, 2 and 3 antigens.
  spinner