Mononucleosis Testing
Thermo Scientific™ Remel™ Color Slide™ II Mononucleosis Test Available on GSA/VA Contract for Federal Government customers only.
Rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
Fisher Healthcare™ Sure-Vue™ Mono Latex Test Kit Available on GSA/VA Contract for Federal Government customers only.
Qualitative and semi-quantitative results
Fisher HealthCare™ Sure-Vue™ Signature Mono Test Kit Available on GSA/VA Contract for Federal Government customers only.
Rapid, lateral flow test, waived for whole blood.
Thermo Scientific™ Remel™ ImmunoDOT™ Mono IgG Test Available on GSA/VA Contract for Federal Government customers only.
An EIA membrane test that simultaneously detects IgG antibodies to EBV, VCA, EBNA, CMV, and Toxoplasma gondii in serum.
Sekisui Diagnostics OSOM™ Mono Test Kit Available on GSA/VA Contract for Federal Government customers only.
Simple, in-office test for the detection of mononucleosis
Quidel™ QuickVue™ Infectious Mononucleosis Test Available on GSA/VA Contract for Federal Government customers only.
Test for whole blood, serum or plasma, cassette with ± format
Alere Clearview™ Mono Test
Provides highly sensitive and specific tests for infectious mononucleosis
HemoCue America Beckman Coulter™ ICON™ Mono Test Kit Available on GSA/VA Contract for Federal Government customers only.
A one-step antibody test for fast, accurate and easy-to-read result at the-point-of care
Meridian Bioscience™ ImmunoCard™ STAT! Mono Test Kit Available on GSA/VA Contract for Federal Government customers only.
Unitary one-step immuno-chromatographic assay for the qualitative detection of infectious mononucleosis heterophile antibodies
Alere Acceava™ Mono Cassette Test
Provides superior performance for the diagnosis of mononucleosis. One-step antibody test for IM that uses direct solid-phase immunoassay technology to detect IM heterophile antibodies in human serum, plasma, or whole blood.
Alere Biostar™ Acceava™ Mono II Kit
Provides superior performance for the diagnosis of mononucleosis. Rapid, flexible procedure reduces office staff hands-on time and follow-up activities.
Polymedco™ Poly stat™ Mono test
Rapid test for visual qualitative detection of heterophile antibodies specific to Infectious Mononucleosis (IM) in human serum, plasma or whole blood
Stanbio™ RELY™ Mono Rapid Test Kit
For qualitative detection of infectious mononucleosis
Alere ColorCard™ Mono Test Available on GSA/VA Contract for Federal Government customers only.
Consists of dyed, color-enhanced horse erythrocytes. Rapid agglutination test intended for use in qualitative and semi-quantitative detection of infectious mononucleosis heterophile antibodies in serum or plasma
Thermo Scientific™ Remel™ GenBio ImmunoDOT™ Mono-M IgM Test Panels
Use with Mono-G Test for serodiagnosis of infectious (EBV) mononucleosis and presumptive serodiagnosis of CMV mononucleosis-like syndrome
Thermo Scientific™ Remel™ ImmunoDOT™ Mono IgM Test Available on GSA/VA Contract for Federal Government customers only.
An EIA membrane test that simultaneously detects the presence of heterophile antibodies and antibodies to EBV-VCA and CMV in serum.
Thermo Scientific™ Remel™ Clearview™ Mono Test
Rapid test used for the qualitative detection of Infectious Mononucleosis (IM) heterophile antibodies in whole blood, serum and plasma.
Thermo Scientific™ Remel™ Mono-Lex System Available on GSA/VA Contract for Federal Government customers only.
Rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.
Alere Inverness Clearview™ Mono-plus II Test
Patented technology provides highly sensitive and specific results for the detection of infectious mononucleos
  spinner