Respiratory Viral Testing
Fisher Healthcare™ Sure-Vue™ RSV Test Kit Available on GSA/VA Contract for Federal Government customers only.
Alternative procedure for bloody and mucoid samples
Abbott™ ID NOW RSV Test Kit
Highly sensitive rapid diagnostic tests for RSV are needed to help guide management in the clinical setting. Combines the benefits of speed and accuracy on an easy to use system.
Thermo Scientific™ Xpect™ RSV Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Respiratory Syncytial Virus (RSV) from nasopharyngeal specimens in neonatal and pediatric patients and get results in 15 minutes.
Thermo Scientific™ BinaxNOW™ RSV CLIA Waived Test Available on GSA/VA Contract for Federal Government customers only.
Detect RSV in 15 minutes with BinaxNOW™ CLIA Waived RSV, a rapid, one-step lateral flow test.
Alere BinaxNOW™ RSV Lateral Flow Test Kits Available on GSA/VA Contract for Federal Government customers only.
Rapid, one-step lateral flow test for Respiratory Syncytial Virus (RSV)
Quidel™ Sofia™ RSV FIA Immunofluorescence Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigens
BD Veritor System RSV (Respiratory Syncytial Virus) Test Kit
Provides objective results on an easy to read digital display, unlike current rapid RSV tests, which requires subjective visual test interpretation to determine positive or negative test result
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel, Respiratory Virus Available on GSA/VA Contract for Federal Government customers only.
An immunofluorescent test used for 7 respiratory viuses: influenza A and B; adenovirus; parainfluenza 1, 2, 3; and RSV in shell vial pre-CPE assays and tube cultures.
Thermo Scientific™ Remel™ Xpect™ RSV Control Kit Available on GSA/VA Contract for Federal Government customers only.
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
Quidel QuickVue™ RSV Test Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid, qualitative detection of RSV antigen directly from nasopharyngeal swab or aspirate specimens from symptomatic pediatric patients
Alere RSV Control Swabs Available on GSA/VA Contract for Federal Government customers only.
For use with BinaxNOW™ RSV Diagnostic test kits
Meridian Bioscience™ TRU RSV™ Tests Available on GSA/VA Contract for Federal Government customers only.
Test under the hood without concern about airflow
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Control Slide Available on GSA/VA Contract for Federal Government customers only.
Remel PathoDx and Respiratory Virus Panel Control Slide is used with PathoDx Respiratory Virus Panel.
Zeptometrix NATtrol™ RP Controls
Designed to evaluate the performance of nucleic acid tests for determination of the presence of bacterial and viral nucleic acids
Zeptometrix NATtrol T.vaginalis Negative Control
Designed to evaluate the performance of nucleic acid test for determination of the presence of T. vaginalis nucleic acids
Alere Clearview™ RSV Test Kit Available on GSA/VA Contract for Federal Government customers only.
Intended for the rapid, qualitative detection of respiratory syncytial virus (RSV) directly from nasopharyngeal swab and nasal aspirate specimens in those under the age of 6 or over the age of 60. Intended for use as an aid in the diagnosis of patients with symptoms consistent with RSV infection.
Zeptometrix NATtrol™ T. vaginalis Positive Control
Designed to evaluate the performance of nucleic acid tests for determination of the presence of T. vaginalis nucleic acids
Meridian Bioscience™ Positive Control
For use with ImmunocardSTAT Flu/RSV
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Parainfluenza 1 Reagent
Remel PathoDx and Respiratory Virus Panel Parainfluenza 1 Reagent is used with PathoDx Respiratory Virus Panel.
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Mounting Fluid
Remel PathoDx and Respiratory Virus Panel Mounting Fluid is used with PathoDx Respiratory Virus Panel.
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Parainfluenza 2 Reagent
Remel PathoDx and Respiratory Virus Panel Parainfluenza 2 Reagent is used with PathoDx Respiratory Virus Panel.
Thermo Scientific™ Remel™ Xpect™ RSV Sample Panel
Remel Xpect RSV Sample Panel is used with Xpect RSV.
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Parainfluenza 3 Reagent
Remel PathoDx and Respiratory Virus Panel Parainfluenza 3 Reagent is used with PathoDx Respiratory Virus Panel.
Thermo Scientific™ Oxoid™ IMAGEN™ Respiratory Virus Screen
Oxoid IMAGEN Respiratory Screen detects RSV, influenza A and B virus, parainfluenza virus 1, 2 and 3 and adenovirus.
Thermo Scientific™ Oxoid™ IMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide
Oxoid IMAGEN Respiratory Syncytial Virus Positive Control is a slide with acetone-fixed human epithelial cells infected with RSV.
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Adenovirus Reagent
Remel PathoDx and Respiratory Virus Panel Adenovirus Reagent is used with PathoDx Respiratory Virus Panel.
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel RSV Reagent
Remel PathoDx and Respiratory Virus Panel RSV Reagent is used with PathoDx Respiratory Virus Panel.
  spinner