Rubella Testing
Fisher Healthcare™ Sure-Vue™ Rubella Latex Test Kit Available on GSA/VA Contract for Federal Government customers only.
Requires a dilution step for 10 IU sensitivity
Fisher Healthcare™ Sure-Vue™ SELECT Rubella Latex Test Available on GSA/VA Contract for Federal Government customers only.
Does not require a dilution step for 10 IU sensitivity
Alere Rubella IgG Microplate Assay
Designed for the detection of IgG antibodies to Rubella virus in serum. The test system is intended to be used to evaluate single sera for immune status or paired sera to demonstrate seroconversion. Flexible incubation times can allow turnaround in 50 to 75 minutes plus wash steps.
Alere Rubella-Plus™ Test Available on GSA/VA Contract for Federal Government customers only.
Simple, two-step procedure
Alere Rubella IgM Microplate Assay
Designed for the detection of IgM antibodies to Rubella virus in serum. The test system is intended to be used to evaluate serologic evidence of active or recent infection with Rubella virus. Flexible incubation times can allow turnaround in 50 to 75 minutes plus wash steps.
Diamedix™ Immunosimplicity™ Rubella IgG Test Kit
Detects IgG antibodies to rubella in human serum
ASI™ Rubella IgM ELISA Test Kit
For the qualitative and semiquantitative detection of human IgM antibodies to Rubella virus
ASI™ Rubella IgG ELISA Test Kit
For the qualitative and semiquantitative detection of human IgG antibodies to Rubella virus
Alere Rubella IgG II Test Kit
Provides increased reliability and run-to-run consistency. Uses substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays. This uniformity of reagents allows for ease-of-automation with microtiter processing systems.
Diamedix™ Immunosimplicity™ Rubella IgM Capture Test Kit
Detects IgM antibodies to rubella in human serum
  spinner