Lysing Agents
eBioscience™ 1-step Fix/Lyse Solution, 10x Non-distribution product as customer accommodation.
Enables lysis of RBCs after staining peripheral blood cells with fluorochrome-conjugated antibodies. Formulated to lyse non-nucleated erythrocytes while maintaining fixed and labeled leukocyte population.
HORIBA ABX Lysebio for ABX Pentra 60, 80 Hematology Analyzer Available on GSA/VA Contract for Federal Government customers only.
For use with ABX Pentra 60, 80 Hematology Analyzer
HORIBA ABX Alphalyse for ABX Micros 60 IM Hematology Analyzer Available on GSA/VA Contract for Federal Government customers only.
Used with ABX Micros 60IM Hematology Analyzer
  spinner