Linearity Calibrators
Streck Cell-Chex™ Cell Body Fluid Control Available on GSA/VA Contract for Federal Government customers only.
Features White Cell Differential Capability
Streck A1c-Cellular™ Linearity Material
For evaluating accuracy and precision of HbA1c procedures over instrument reportable range
Streck CD-Chex™ CD34 Available on GSA/VA Contract for Federal Government customers only.
Positive procedural control for enumerating CD34 positive cells by flow cytometry
Streck CD-Chex TdT™ Plus Assay
Quality control material for evaluating intracellular and surface antigens including CD1a, CD2, CD4, CD5, CD7, CD8, CD34, cytoplasmic CD3 and nuclear TdT (Terminal deoxynucleotidyl transferase)
Streck Retic-Chex™ Linearity
For calibration verification assessment, also for establishing the analytic linearity of automated reticulocyte counting instruments
Streck Cell-Chex™ Auto Body Fluid Cell Count Control Available on GSA/VA Contract for Federal Government customers only.
Eliminate the need for manual hemacytometer testing
Streck Cell-Chex™ Auto Body Fluid Cell Count Control
Three-level spinal and body fluid control for evaluating accuracy and precision of hematology instruments that measure blood cell counts in patient body fluid samples
Streck LBC-Chex™ Lamellar Body Count Control
For use on automated hematology analyzers
  spinner