Urine Testing Reagents, Controls and Strips
Roche Diagnostics Chemstrip™ 10UA Urine Test Strips
Reagent strips for use with Roche Diagnostics Chemstrip 101 Urine Analyzer and Chemstrip Criterion™ II Urine Analyzer
Roche Diagnostics POC Chemstrip™ Urine Test Strips
Accuracy, performance and testing simplicity
Fisherbrand™ Urine Reagent Strips
Read visually or use with the AimStrip™ Urine Analyzer
Fisherbrand™ Urine Sedimentation System Available on GSA/VA Contract for Federal Government customers only.
Designed to produce the same results as the most popular style urine sedimentation system using the same test procedure
Siemens Healthineers Multistix™ and Uristix™ Reagent Strips Available on GSA/VA Contract for Federal Government customers only.
Visually and instrumentally readable urinalysis strips
KOVA™ Glasstic™ Slide 10 Available on GSA/VA Contract for Federal Government customers only.
Precision manufactured, 10-chambered slide available without a hemocytometer-type grid
Siemens Healthineers Urinalysis Reagent Test Strips Available on GSA/VA Contract for Federal Government customers only.
Single-use, plastic strips with multiple reagent test areas
Alere E. histolytica II Test Kit Available on GSA/VA Contract for Federal Government customers only.
State-of-the art ELISA test for the detection of Entamoeba histolytica pathogens in fecal specimens. Uses substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays. This uniformity of reagents allows for ease-of-automation with microtiter processing systems.
KOVA™ Glasstic™ Slide 10 with Grids Available on GSA/VA Contract for Federal Government customers only.
Use optically clear plastic for microscopic analysis with same refractive index as glass.  Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
Alere E. HISTOLYTICA QUIK CHEK™
The E. HISTOLYTICA QUIK CHEK™ test is a rapid test for the qualitative detection of Entamoeba histolytica in fecal samples and targets an adhesin unique to E. histolytica. The test can be used for quick and reliable diagnosis of amebiasis.
Germaine™ Laboratories AimTab™ Ketone Tablets
For the semi-quantitative determination of Ketones (acetoacetic acid & acetone) in urine, serum, plasma and whole blood. Replaces the discountinued Bayer Acetest. Same test procedure at the Acetest™. Detects as little as 5mg/dl of acetoacetic acid in urine.
Meridian Bioscience™ Para-Pak SpinCon™ Concentration System Available on GSA/VA Contract for Federal Government customers only.
Requires only one spin for recovery of protozoan cysts, trophs and helminth ova and larvae
PTS Diagnostics PTS PANELS™ Lipid Panel Test Strips
Quick and easy measurements in whole blood using CardioChek™ PA Analyzer
Germaine™ Laboratories AimTab™ Reducing Substances Tablets
For the semi-quantitative determination of the amount of reducing substances (glucose, lactose, fructose, galactose, pentoses) in urine, stool, wine, or any other liquids. Tablets provides clinically useful information on carbohydrate metabolism. FOR FORENSIC USE ONLY.
Alere Instant Bluing Tablets
Prevent adulteration prior to specimen collection
Germaine™ Laboratories AimTab™ Reducing Substances Tablets
Germaine™ Laboratories AimTab™ Reducing Substances Tablets are for the semi-quantitative determination for the amount of reducing substances (generally glucose) in urine and other fluids
KOVA™ Stain Available on GSA/VA Contract for Federal Government customers only.
Modified Sternheimer-Malbin stain that facilitates differentiation and enhances visualization of formed elements in urine sediment
KOVA™ Glasstic™ Slide II Available on GSA/VA Contract for Federal Government customers only.
Four-chambered slide
Ascensia Diabetes Care Urinalysis Reagent Test Strips Available on GSA/VA Contract for Federal Government customers only.
Single-use, plastic strips with multiple reagent test areas
Roche Diagnostics Chemstrip™ UA Calibration Strips
For use with Roche Diagnostics™ Chemstrip 101 Urine Analyzer and Chemstrip Criterion™ II Urine Analyzer
Oak Ridge Urine Sediment Tubes
Designed for use with Sysmex™ and Siemens™ Atlas™ instruments
Siemens Healthineers Multistix PRO™ Reagent Strips Available on GSA/VA Contract for Federal Government customers only.
Proprietary dip-and-read tests, Protein Low (Albumin), Protein High, and Creatinine
  spinner