Test Strips
Alere E. histolytica II Test Kit Available on GSA/VA Contract for Federal Government customers only.
State-of-the art ELISA test for the detection of Entamoeba histolytica pathogens in fecal specimens. Uses substrate, stop solution, and wash buffer that are interchangeable with other Wampole assays. This uniformity of reagents allows for ease-of-automation with microtiter processing systems.
Alere E. HISTOLYTICA QUIK CHEK™
The E. HISTOLYTICA QUIK CHEK™ test is a rapid test for the qualitative detection of Entamoeba histolytica in fecal samples and targets an adhesin unique to E. histolytica. The test can be used for quick and reliable diagnosis of amebiasis.
Meridian Bioscience™ Para-Pak SpinCon™ Concentration System Available on GSA/VA Contract for Federal Government customers only.
Requires only one spin for recovery of protozoan cysts, trophs and helminth ova and larvae
PTS Diagnostics PTS PANELS™ Lipid Panel Test Strips
Quick and easy measurements in whole blood using CardioChek™ PA Analyzer
PTS Diagnostics PTS PANELS™ Triglycerides Test Strips
Quick and easy measurement of triglycerides in whole blood using CardioChek™ PA Analyzer for professional and home use
Roche Diagnostics Glucose Test Strips for Accutrend Plus™ Meters
For the determination of glucose in capillary whole blood
PTS Diagnostics PTS PANELS™ CHOL+HDL Panel Test Strips
Quick and easy measurement of total cholesterol and HDL cholesterol in whole blood using CardioChek™ PA analyzer
PTS Diagnostics PTS PANELS™ CHOL+GLU Panel Test Strips
Quick and easy measurement of cholesterol and glucose in whole blood using CardioChek™ PA Analyzer for home and professional use
PTS Diagnostics PTS PANELS™ Cholesterol Test Strips
Quick and easy measurement of cholesterol in whole blood using CardioChek™ PA Analyzer for professional and home use
PTS Diagnostics PTS PANELS™ CHOL+HDL+GLU Panel Test Strips
Quick and easy measurement of cholesterol, HDL cholesterol and glucose in whole blood using CardioChek™ PA Analyzer
PTS Diagnostics PTS PANELS™ HDL Cholesterol Test Strips
Quick and easy measurement of HDL Cholesterol from whole blood using CardioChek™ PA Analyzer for professional and home use
PTS Diagnostics PTS PANELS™ Glucose Test Strips
Quick and easy measurement of glucose in whole blood using CardioChek™ PA Analyzer for professional and home use
LifeSign Status BTA Urine Test Kit
Status BTA is a rapid test for the qualitative detection of bladder tumor associated antigen in human urine.
Cen-Med Parasitology Stool Transport Kit
Vials are used in the routine collection, preservation, and transportation, of stool specimens
Meridian Bioscience™ Para-Pak™ Stool Concentrator System Lab Supplies
General laboratory supplies for use in processing of stool samples
Meridian Bioscience™ Para-Pak™ Macro-CON™ Accessories
For use in Macro-CON concentration of eggs, larvae, and cysts
  spinner