Urine Sediment Testing
Fisherbrand™ Urine Sedimentation System Available on GSA/VA Contract for Federal Government customers only.
Designed to produce the same results as the most popular style urine sedimentation system using the same test procedure
KOVA™ Glasstic™ Slide 10 Available on GSA/VA Contract for Federal Government customers only.
Precision manufactured, 10-chambered slide available without a hemocytometer-type grid
KOVA™ Glasstic™ Slide 10 with Grids Available on GSA/VA Contract for Federal Government customers only.
Use optically clear plastic for microscopic analysis with same refractive index as glass.  Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
KOVA™ Stain Available on GSA/VA Contract for Federal Government customers only.
Modified Sternheimer-Malbin stain that facilitates differentiation and enhances visualization of formed elements in urine sediment
KOVA™ Glasstic™ Slide II Available on GSA/VA Contract for Federal Government customers only.
Four-chambered slide
Oak Ridge Urine Sediment Tubes
Designed for use with Sysmex™ and Siemens™ Atlas™ instruments
Eng Scientific Urinary Sediment Stain
A quick staining procedure by which a drop of stain solution is directly added to urine sediment
Globe Scientific Quick-Prep™ Urinalysis Kit
Tube and pipette system that isolates 1mL of urine sediment for microscopic examination
KOVA™ Decanting Rack
Ideal for KOVA Super Tubes and Enonomy Tubes
  spinner