Hemacytometers
MilliporeSigma™ Scepter 2.0 Cytometer Sensors Available on GSA/VA Contract for Federal Government customers only.
Designed for use with the Scepter 2.0 Automated Handheld Cell Counter (cat. no. PHCC20040). MilliporeSigma™ Scepter 2.0 Cytometer Sensors ensure accurate, reproducible counts and eliminate pipetting errors and uneven sample loading. Available in two apertures to count cells between 4 and 25 μm.
SKC, Inc. C-Chip™ Disposable Hemacytometers
A significant improvement over conventional glass hemacytometers
Hausser Scientific Bright-Line™ Counting Chamber
Diamond etched with double improved Neubauer ruling
Hausser™ Bright-Line™ Phase Hemacytometer Available on GSA/VA Contract for Federal Government customers only.
Counting chamber with reduced thickness for use on phase microscopes
Hausser Scientific™ Bright-Line™ and Hy-Lite™ Counting Chambers Available on GSA/VA Contract for Federal Government customers only.
For accurate white blood cell, red blood cell, and platelet counts
Hausser Scientific Bright Line™ Counting Chamber Available on GSA/VA Contract for Federal Government customers only.
Counting Chamber; Bright Line; Hausser Scientific; Molded Pyrex glass; Double improved Neubauer ruling
Hausser Scientific™ Petroff-Bacteria Counter
Hausser Scientific Petroff-Hausser Bacteria Counter features improved Neubauer ruling on single plateau and is popular for bacteria and sperm counting.
Millennium Sciences CELL-VU™ Disposable Sperm Count Cytometers
Cytometers, Sperm Count; CELL-VU; For undiluted specimens; Disposable; 3 x 1 in. glass slide and coverslip w/counting grid etched on surface; Slide has 2 chambers for dual determinations; 25 slides and 50 cover slips/box
Hausser Scientific™ Levy™ Hemacytometer Chamber Set
This hemacytometer has an etched field with an opaque-pattern Levy chamber and double Neubauer ruling
Millennium Sciences™ CELL-VU™ CBC Disposable Hemacytometers
Ideal for manual erythrocyte, leukocyte, platelet, and eosinophil cell counts, the disposable CELL-VU Hemacytometer consists of a standard (3 × 1 in.) specially designed glass slide and coverslip
Hausser Scientific™ Hy-Lite™ Counting Chamber Set
Supplied with polyethylene carrying case
Hausser Scientific™ Bright-Line™ Counting Chamber Set
Supplied in polyethylene carrying case
Hausser Scientific™ Nageotte Bright-Line™ Hemacytometer Available on GSA/VA Contract for Federal Government customers only.
Manually count low leukocyte concentrations below the capabilities of a standard hemacytometer. Hausser Nageotte Bright-Line Hemacytometer can accurately measure down to 0.1 cell/mL.
MilliporeSigma™ Scepter 2.0 Automated Handheld Cytometer Accessory: O-Ring Kit Available on GSA/VA Contract for Federal Government customers only.
For use with MilliporeSigma™ Scepter 2.0 (see cat. no. PHCC20040)
MilliporeSigma™ Scepter 2.0 Automated Handheld Cytometer Accessory: USB Cable
For use with MilliporeSigma™ Scepter 2.0 (see cat. no. PHCC20040)
Hausser Scientific Howard Mold Counting Chamber
Designed for use in determining mold counts
MilliporeSigma™ Scepter 2.0 Automated Handheld Cytometer Accessory: Test Beads
For use with MilliporeSigma™ Scepter 2.0 (see cat. no. PHCC20040)
MilliporeSigma™ Scepter 2.0 Automated Handheld Cytometer Accessory: Universal Power Adapter Available on GSA/VA Contract for Federal Government customers only.
For use with MilliporeSigma™ Scepter 2.0 (see cat. no. PHCC20040)
MilliporeSigma™ Scepter 2.0 Automated Handheld Cytometer Available on GSA/VA Contract for Federal Government customers only.
Cell counting in a portable, personalized format
Hausser Scientific™ Hemocytometer Products for Science Education
Get quick, accurate readings of red and white blood cells.
  spinner