Cardiac Testing
Quidel Triage MeterPro BNP Panel 25/PK
Triage BNP is a rapid, point-of-care CLIA-Waived fluorescence immunoassay used with the Triage MeterPro. It is used to measure B-type natriuretic peptide (BNP) in EDTA anticoagulated whole blood or plasma specimens.
Quidel Triage MeterPro Cardiac Panel 25/PK
Triage Cardiac Panel is a rapid, point of care fluorescence immunoassay for use on the Triage MeterPro. This test is designed to determine the level of creatine kinase MB (CK-MB), myoglobin and troponin I in EDTA anticoagulated whole blood or plasma specimens.
Quidel Triage MeterPro Cardiac Panel Troponin/CK-MB 25/PK
Triage Cardiac Panel is a rapid, point of care fluorescence immunoassay for use on the Triage MeterPro. This test is designed to determine the level of creatine kinase MB (CK-MB) and troponin I in EDTA anticoagulated whole blood or plasma specimens.
Quidel Triage MeterPro Cardiac Panel Troponin 25/PK
Triage Cardiac Panel is a rapid, point of care fluorescence immunoassay for use on the Triage MeterPro. This test is designed to determine the level of troponin I in EDTA anticoagulated whole blood or plasma specimens. This can be used as an aid in the diagnosis of myocardial infarction (injury).
Tosoh Bioscience AIA-PACK™ Cardiac Markers Assays: Assay Test Cups
Pre-measured, single unit-dosed dry reagents
Tosoh Bioscience AIA-PACK™ Cardiac Markers Assays: Calibrators
Pre-measured, single unit-dosed dry reagents
LifeSign Lifesign MI™ Cardiac Markers Test Kit
Lifesign MI™ tests are for rapid detection of cardiac markers
Tosoh Bioscience AIA-PACK™ Cardiac Markers Assays: Solutions
Pre-measured, single unit-dosed dry reagents
PTS Diagnostics Accessories for CardioChek™ PA Analyzer
Experience increased versatility and usefulness with these accessories
Quidel Triage MeterPro Combo Cardiac Panel, BNP, D-DIMER 75 total tests
See Cardiac Panel, BNP Test and D-Dimer Test for more detail
Quidel Triage MeterPro Cardiac Troponin only plus BNP 25/PK
See Cardiac Panel (97021HS), and BNP Test for more detail
Quidel Triage MeterPro Combo Troponin, Troponin/CK-MB with BNP, and D-Dimer 75 total tests
See Cardiac Panel (97022HS), BNP Test and D-Dimer Test for more detail
PTS Diagnostics CardioChek™ PA Analyzer
Cost-efficient, user-friendly diagnostic point-of-care analyzer, which can accurately determine multiple patient values
Quidel Triage MeterPro Combo Cardiac Panel/BNP 50 total tests
See Cardiac Panel and BNP Test for more detail
Quidel Triage MeterPro Combo Troponin, BNP, D-Dimer 75 total tests
See Cardiac Panel (97021HS), BNP Test and D-Dimer Test for more detail
Quidel Triage MeterPro Combo Troponin/CK-MB and BNP 50 total tests
See Cardiac Panel (97022HS) and BNP Test for more detail
Diazyme Laboratories Enzymatic Homocysteine 5 Level Calibrator
Diazyme 5 Level Homocysteine Calibrator Set is for the testing of Homocysteine in serum.
Diazyme Laboratories Apoliprotein B Assay
Diazyme Apolipoprotein B Assay is intended for the quantitative in vitro determination of Apolipoprotein B (Apo B) in serum.
Diazyme Laboratories Lipoprotein (a) Calibrator
Diazyme Lipoprotein (a) Calibrator set is for the testing of Lipoprotein (a) in samples of human serum.
Diazyme Laboratories Cardiac Troponin I Calibrator
Diazyme Cardiac Tropoinin I Calibrator set is for the testing of Cardiac Troponin I in human serum.
Diazyme Laboratories Apolipoprotein A-1/B Calibrator
Diazyme Apolipoprotein A-1/B Calibrator set is for the testing of Apolipoprotein A-1/B in human serum.
Diazyme Laboratories Apolipoprotein A-1 Assay
Diazyme Apolipoprotein A-I Assay is intended for the in vitro quantitative determination of Apolipoprotein A-I (Apo A-I) in serum.
Diazyme Laboratories HDL/LDL Calibrator
Diazyme HDL/LDL Calibrator set is for the testing of high density lipoprotein cholesterol and low-density lipoprotein cholesterol in samples of human serum.
Diazyme Laboratories Cardiac Troponin I Assay
Diazyme Cardiac Troponin I Assay is for the determination of cardiac troponin I in serum or plasma by latex enhanced immunoturbidimetric method.
Diazyme Laboratories PLAC™ Test for Lp-PLA2
Enzyme assay for the quantitative determination of Lp-PLA2 activity in human plasma and serum.
Vernier Vernier EKG Electrodes Products for Science Education
Replacement package of 100 electrodes for use with EKG sensor.
  spinner