FOBT Testing
Quidel™ QuickVue™ iFOB Test Available on GSA/VA Contract for Federal Government customers only.
Immunochemical FOB test—a more analytically sensitive alternative to Guiac methods
Hemosure™ iFOB Test Kits: Cassettes Only
Cassettes for use with hemosure iFOB Test Kits
Fisher Healthcare™ Sure-Vue™ Signature™ Immunochemical Fecal Occult Blood Test (iFOBT)
Immunochemical device for the rapid qualitative determination of fecal occult blood. The Fisher Healthcare Sure-Vue™ Signature™ (iFOBT) can be performed in a laboratory, physician’s office and is useful in detecting gastrointestinal (GI) bleeding from a number of disorders.
Hemosure™ iFOB Test Kits
Qualitative, one-step, odorless iFOB test with high sensivity and specificity
Fisher Healthcare™ Sure-Vue™ Fecal Occult Blood Slide Test System Available on GSA/VA Contract for Federal Government customers only.
Available in traditional guaiac format for routine screening applications and extra-sensitive format for more stringent testing
HemoCue America Beckman Coulter™ Hemoccult™ SENSA™ Fecal Occult Blood Slide Test System Available on GSA/VA Contract for Federal Government customers only.
Provides enhanced detection at low, but abnormal, levels of hemoglobin
Hemosure™ iFOB Test Kits: Home Kit Mailers
For use with Hemosure iFOB Test Kits
Hemosure™ iFOB Test Kits: Collection Tubes
For use with Hemosure iFOB Test Kits
HemoCue America Beckman Coulter™ Hemoccult™ Fecal Occult Blood Slide Test System Available on GSA/VA Contract for Federal Government customers only.
Several packaging configurations are available for different testing needs
Sekisui Diagnostics OSOM™ iFOB Test
OSOM iFOBT gives the physician and the patient a simpler, more compliant cancer screening option that is highly accurate (50 ng hHb/mL) and specific to human hemoglobin
Aerscher Diagnostics HemaPrompt™ Blood Test Kit
Guaiac-based combination occult blood test for fast and reliable results
Immunostics Hema-Screen SPECIFIC™ iFOBT Complete Test Kit
Use for the rapid and qualitative determination of human blood in fecal sample. Innovative immunochemical collection system uses a single iFOBT cassette and a single patented Devel-A-Tab™ sampling card to detect human hemoglobin [Hb (globin)] in fecal stool specimens.
Hemosure™ iFOB Test Kits: External Control Sets
For use with Hemosure iFOB Test Kits
Immunostics Hema-Screen™ SPECIFIC™ iFOBT Cassette Kit
Use for the rapid and qualitative determination of human hemoglobin in fecal samples by immunochemical methods. This kit for direct patient sampling uses a collection tube and iFOBT cassette.
Helena Laboratories™ ColoScreen™ FOB Test Kit Available on GSA/VA Contract for Federal Government customers only.
Convenient triple slide provides three successive tests in one unit
Polymedco™ Accessories for OC-Light iFOBT Test Kits
Collection of accessories to supplement OC-Light iFOBT kits
HemoCue America Beckman Coulter™ Hemoccult™ Fecal Occult Blood Slide Test System: Accessories Available on GSA/VA Contract for Federal Government customers only.
Accessories for Beckman Coulter Hemoccult Fecal Occult Blood Slide Test System (cat. no. SK60151)
Siemens Healthineers Hema-Chek™ Fecal Occult Blood Test Slides and Developer Available on GSA/VA Contract for Federal Government customers only.
This simple, economical test produces results in 30 seconds.
Immunostics Hema-Screen™ Fecal Occult Blood Test Kit
For detecting occult blood in stool
Cambridge Diagnostic Products, Inc.™ Camco™ Guaiac Tablets
For qualitative determination of fecal occult blood
Immunostics Hema-Screen™ STAT FOBT Test
Use for the rapid, convenient and qualitative detection of asymptomatic occult bleeding.
Immunostics Hema-Screen SPECIFIC™ iFOBT Controls
External controls utilizing human hemoglobin to aid end user in verifying functionality of the immunoassay
HemoCue America Beckman Coulter™ Hemoccult™ ICT 2-Day Patient Screening Kit
Immunochemical FOBT (iFOBT) with high specificity and sensitivity
Stanbio™ Hema-Screen™ Lab Pack
Designed to collect specimens and perform assay in laboratory
  spinner