Influenza Testing
Thermo Scientific™ Xpect™ Flu A & B Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens, and get results in 15 minutes.
BD Veritor™ Plus Analyzer
Point of Care detection of infectious diseases such as influenza A and B, respiratory syncytial virus (RSV) and group A strep. BD Veritor Plus Analyzer provides lab-quality immunoassay test results within minutes.
Abbott™ ID NOW Influenza A and B 2 Edge
The fastest molecular flu test just got faster and even better. Now with early detection and positive results in as few as 5 minutes, allowing you to make effective clinical decisions sooner. Abbott ID NOW is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.
Quidel Sofia™ Influenza A+B FIA Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid differential diagnosis of acute influenza A and influenza B viral infections
BD Veritor™ System for the Rapid Detection of Flu A+B Edge
Unique adaptive-read technology helps reduce false-positive results
Quidel QuickVue™ Influenza A+B Test Available on GSA/VA Contract for Federal Government customers only.
Aids in the rapid differential diagnosis of acute influenza A and B virus infection
Sekisui Diagnostics OSOM™ Ultra Flu A and B Rapid Test Edge
OSOM Ultra Flu A&B is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens
Quidel Solana™ Flu Assay Startup Kit Available on GSA/VA Contract for Federal Government customers only.
For rapid differential diagnosis of acute influenza A and influenza B viral infections.
Meridian Bioscience™ ImmunoCard STAT!™ FLU A&B
Meridian Bioscience ImmunoCard™ STAT! FLU A&B is a rapid Flu test for Nasal and NP swabs
Sekisui Diagnostics Silaris™ Influenza A&B Test Kit, 25/PK
The CLIA Waived Silaris™ Influenza A&B Test performed on the Silaris Dock is a molecular in vitro diagnostic test utilizing (PCR) and lateral flow technology for the qualitative, visual detection and differentiation of influenza A&B.
ZeptoMetrix NATtrol™ Influenza/RSV Negative Control
Designed for use in evaluating and monitoring the performance of nucleic acid tests for determination of the presence of respiratory virus nucleic acids
BD Veritor™ Plus Analyzer with Flu A+B CLIA Waived Flu Assay Kits (2 x 30)
Point of Care lab-quality detection of influenza A and B. BD Veritor™ Plus Analyzer with Flu A+B CLIA Waived Flu Assay Kits (2 x 30) includes an advanced chromatographic instrument and two immunoassay kits to detect antigens from respiratory specimens of symptomatic patients in 10 minutes.
ZeptoMetrix NATtrol™ Influenza A/B Positive Control
Designed for use in evaluating and monitoring the performance of nucleic acid tests for determination of the presence of respiratory virus nucleic acids
Thermo Scientific™ Remel™ Xpect™ Flu A and B Control Swab Available on GSA/VA Contract for Federal Government customers only.
Remel Xpect and Flu A & B Control Swabs are used as external quality control material for the Xpect Flu A and B test.
Abbott™ ID NOW Influenza A and B Control Swab Kit Available on GSA/VA Contract for Federal Government customers only.
Provides molecular results in less than 15 minutes, allowing you to make effective clinical decisions sooner. Abbott ID NOW is a rapid, instrument-based system for the qualitative detection of infectious diseases using isothermal nucleic acid amplification technology.
ZeptoMetrix NATtrol™ Influenza External Run Controls Available on GSA/VA Contract for Federal Government customers only.
Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable
Abbott™ BinaxNOW™ Nasopharyngeal Swab Accessory Pack Available on GSA/VA Contract for Federal Government customers only.
Swabs and transport vials for use with BinaxNOW Diagnostic test kits
Sekisui Diagnostics Silaris™ Influenza A&B Control  Kit
The Silaris™ Influenza  A&B Control  Kit is intended for in vitro diagnostic use in quality control testing with the Silaris Influenza A&B Test.
Meridian Bioscience™ ImmunoCard STAT!™ FLU A&B Accessory Kit
Meridian Bioscience ImmunoCard™ STAT! FLU A&B Accessory Kit for testing nasal aspirate/washes and samples in transport media
Thermo Scientific™ Oxoid™ IMAGEN™ Parainfluenza Group
Oxoid IMAGEN Parainfluenza Group detects and confirms parainfluenza virus antigens in nasopharyngeal aspirates and cell culture preparations.
Thermo Scientific™ Oxoid™ IMAGEN™ Parainfluenza Typing
Oxoid IMAGEN Parainfluenza Typing detects and differentiates Parainfluenza virus Types 1, 2 and 3 antigens.
Fisherbrand™ Sure-Vue™ Signature Test Influenza A&B Kit
Delivers rapid detection of influenza type A and B nucleoprotein antigens directly from nasal and nasopharyngeal swab specimens from patients. High sensitivity and specificity allow for accurate diagnostic testing that is both efficient and cost-effective
Thermo Scientific™ Wellcogen™ Haemophilus influenzae b Rapid Latex Agglutination Test Available on GSA/VA Contract for Federal Government customers only.
Rapidly detect antigens to Haemophilus influenzae present in human body fluids or blood cultures by latex agglutination.
Thermo Scientific™ Remel™ Xpect™ Flu A & B Sample Panel
Remel Xpect & Flu A and B Sample Panel is used with Xpect Flu A & B test kit.
Thermo Scientific™ Oxoid™ IMAGEN™ Influenza A & B Available on GSA/VA Contract for Federal Government customers only.
For the detection and identification of Influenza virus strains A and B in clinical specimens or cell cultures
BD Veritor™ Plus Analyzer with Flu A+B CLIA Moderate Complex Flu Assay Kits (2 x 30)
Point of Care lab-quality detection of influenza A and B. BD Veritor™ Plus Analyzer with Flu A+B CLIA Moderate Complex Flu Assay Kits (2 x 30) includes an advanced chromatographic instrument and two immunoassay kits to detect antigens from respiratory specimens of symptomatic patients in 10 min.
Abbott™ Flu Starter Kit
Flu Starter Kit combines the DIGIVAL Complete Kit (Catalog #23046680) with Influenza A and B Test Cards (Catalog #23046682) and an optional thermal printer (Catalog #23046687) for all-in-one influenza testing
Sekisui Diagnostics OSOM™ Ultra Flu A&B Test Control Swab Kit
OSOM Ultra Flu A&B is a rapid qualitative test that detects influenza type A and type B antigens directly from nasal swab, nasopharyngeal swab and nasal aspirate/wash specimens.
  spinner