Hematology Stains
Fisher HealthCare™ PROTOCOL™ Hema 3™ Manual Staining System and Stat Pack Available on GSA/VA Contract for Federal Government customers only.
For 30-second, three-step staining comparable to the Wright-Giemsa method
Fisher HealthCare™ PROTOCOL™ Hema 3™ Fixative and Solutions Available on GSA/VA Contract for Federal Government customers only.
For 30-second, three-step staining comparable to the Wright-Giemsa method
Fisher HealthCare™ PROTOCOL™ Hema-G Stains for Geometric Data Hemastainer™ Available on GSA/VA Contract for Federal Government customers only.
Quality-control tested on human blood smears to ensure proper staining and absence of precipitate
Fisher HealthCare™ PROTOCOL™ Stains for HEMA-TEK™ Automated Slide Stainers Available on GSA/VA Contract for Federal Government customers only.
Allow varied staining times to suit individual preferences
EMD Millipore™ Harleco™ Wright Stain Solutions Available on GSA/VA Contract for Federal Government customers only.
For use with the MIDAS™ III Automated Stainer
EMD Millipore™ Harleco™ Azure B Giemsa Stain Solution Available on GSA/VA Contract for Federal Government customers only.
For use with the MIDAS™ III Automated Stainer
Sigma Diagnostics™ Wright Stain, Modified
Used for differentially staining the cellular elements of blood
Cambridge Diagnostic Products, Inc.™ Camco Quik Stain™ and Quik Stain™ II Available on GSA/VA Contract for Federal Government customers only.
Single, easy-to-use solutions provide fast and consistent differential staining of all cell types in less than 30 seconds
EMD Millipore™ Harleco™ Giemsa Stain Solution Available on GSA/VA Contract for Federal Government customers only.
For use with the MIDAS™ III Automated Stainer
EMD Millipore™ HARLECO™ Giemsa Stain Available on GSA/VA Contract for Federal Government customers only.
For red blood cell morphology and white blood cell differentiation
Sigma Diagnostics™ Phosphate buffer pH 7.2 at 25°C
For use as a buffer in various Romanowsky type staining procedures
Sigma Diagnostics™ Wright-Giemsa Stain, Modified
Used in staining blood or bone marrow films
Fisher HealthCare™ PROTOCOL™ Hemaspray™ Reagents for the Wescor Aerospray™ Available on GSA/VA Contract for Federal Government customers only.
Manufactured using a two-part aqueous Romanowsky stain formulation
Cambridge Diagnostic Products, Inc.™ Camco™ Wright Stain, Fixative and Buffer
Plastic container is easy to store and allows convenient dispensing into slide stainer traps
Astral Diagnostics™ Quick III™ Set
A three-step rapid differential stain for the differential staining of blood smears, blood parasites, microbiological organisms, and bone marrow specimens
Sigma Diagnostics™ Iron Stain Kit
For histologic staining of iron in blood or bone marrow films
EMD Millipore™ Harleco™ May-Grunwald Solution Available on GSA/VA Contract for Federal Government customers only.
For use with the MIDAS™ III Automated Stainer
Eng Scientific Stat-Quick Wright and Wright Giemsa Stain Kits
Utilized in both stat and routine laboratories
EMD Millipore™ Harleco™ Hemacolor™ Solution I Available on GSA/VA Contract for Federal Government customers only.
Used as fixative for all Wright and Wright-Giemsa staining protocols
Sigma Diagnostics™ Giemsa stain, modified
Used in staining blood or bone marrow films
EMD Millipore™ Harleco™ APHA Wright Giemsa Solution Available on GSA/VA Contract for Federal Government customers only.
For use with the MIDAS™ III Automated Stainer
Sigma Diagnostics™ Fetal Hemoglobin Kit
For acid elution, semi-quantitative determination of fetal hemoglobin in blood smears
EMD Millipore™ Harleco™ Hemacolor™ Solution III Available on GSA/VA Contract for Federal Government customers only.
Replacement for methylene blue component of Hemacolor™ Stain Set
Eng Scientific CSF Diluting Fluid
Easily differentiate red blood cells from white blood cells with this diluting fluid that hemolyzes all red cells and stains white cells.
Sigma Diagnostics™ May-Grunwald Stain
For differential staining of cellular elements of blood
  spinner