Autopsy Sinks
Mopec HF200 Organ Rinse Dissecting Station with Large Sink Available on GSA/VA Contract for Federal Government customers only.
Allow for multiple dissecting boards to be placed next to each other directly in front of the subdivided long rinse sink. Mopec™ HF200 Organ Rinse Dissecting Station with Large Sink includes eight removable dividers to allow the separation of organs during the rinsing process.
Mopec Right Hand Approach Elevating Wall Mount Autopsy Sink Available on GSA/VA Contract for Federal Government customers only.
Designed to provide an adjustable working area ergonomically correct for users of various heights. Mopec™ Right Hand Approach Elevating Wall Mount Autopsy Sink can be surface mounted or recessed within the thickness of the wall.
Mopec Embalming Cart with 27 Inch Tray Available on GSA/VA Contract for Federal Government customers only.
Utilizes roller type body storage and processing equipment typically used for most anatomy departments and high capacity embalming facilities. Mopec™ Embalming Cart with 27 Inch Tray incorporates two full length plenum exhaust chambers to fully ventilate the cadaver during the embalming.
Mopec Double Faucet Autopsy Scrub Sink Available on GSA/VA Contract for Federal Government customers only.
Essential product for any anatomy lab providing a specific place to wash hands before and after an autopsy or any handling of infectious material. Mopec™ Double Faucet Autopsy Scrub Sink has no parting seams which can potentially collect bacteria.
Mopec Left Approach Wall Mount Autopsy Sink Available on GSA/VA Contract for Federal Government customers only.
Save time and space with a wall mounted autopsy disssection sink. Mopec™ Left Approach Wall Mount Autopsy Sink features cart trays that overhang the sink to allow for proper fluid drainage.
Mopec Autopsy Tank Service Sink
Accommodates all Mopec autopsy carts and includes most of the amenities to perform an autopsy. Mopec™ Autopsy Tank Service Sink is a pedestal style autopsy service island that features back draft ventilation.
Mopec HF100 Organ Rinse Dissecting Station with Large Sink Available on GSA/VA Contract for Federal Government customers only.
Allow for multiple dissecting boards to be placed next to each other directly in front of the subdivided long rinse sink. Mopec™ HF100 Organ Rinse Dissecting Station with Large Sink includes eight removable dividers to allow the separation of organs during the rinsing process.
Mopec Dual Approach Wall Mount Autopsy Sink Available on GSA/VA Contract for Federal Government customers only.
Save time and space with wall mounted autopsy dissection sink. Mopec™ Dual Approach Wall Mount Autopsy Sink features a cart tray that hangs over the sink to allow for proper fluid drainage.
Mopec FD200 Embalming Sink Available on GSA/VA Contract for Federal Government customers only.
Complete with an extra long sink and an extra tall faucet with built in sterilization that is good for trocars. Mopec™ FD200 Embalming Sink is constructed of all stainless steel.
Mopec Right Approach Wall Mount Autopsy Sink Available on GSA/VA Contract for Federal Government customers only.
Save time and space with a wall mounted autopsy disssection sink. Mopec™ Right Approach Wall Mount Autopsy Sink features cart trays that overhang the sink to allow for proper fluid drainage.
Mopec Embalming Cart with 23 Inch Tray Available on GSA/VA Contract for Federal Government customers only.
Utilizes roller type body storage and processing equipment typically used for most anatomy departments and high capacity embalming facilities. Mopec™ Embalming Cart with 23 Inch Tray incorporates two full length plenum exhaust chambers to fully ventilate the cadaver during the embalming.
Mopec Single Faucet Autopsy Scrub Sink Available on GSA/VA Contract for Federal Government customers only.
Essential product for any anatomy lab providing a specific place to wash hands before and after an autopsy or any handling of infectious material. Mopec™ Single Faucet Autopsy Scrub Sink has no parting seams which can potentially collect bacteria.
Mopec Elevating Center Approach Wall Mount Autopsy Sink Available on GSA/VA Contract for Federal Government customers only.
Designed to provide an adjustable working area ergonomically correct for users of various heights. Mopec™ Elevating Center Approach Wall Mount Autopsy Sink can be surface mounted or recessed within the thickness of the wall.
Mopec Center Approach Embalming Sink Available on GSA/VA Contract for Federal Government customers only.
Incorporates a body rinse fixture for the cleaning and rinsing process during the final stages of body embalming. Mopec™ Center Approach Embalming Sing possesses all the necessary features for full body embalming.
Mopec Center Approach Wall Mount Autopsy Sink Available on GSA/VA Contract for Federal Government customers only.
Save time and space with wall mounted autopsy dissection sink. Mopec™ Center Approach Wall Mount Autopsy Sink features a cart tray that hangs over the sink to allow for proper fluid drainage.
Mopec Left Approach Embalming Sink Available on GSA/VA Contract for Federal Government customers only.
Incorporates a body rinse fixture for the cleaning and rinsing process during the final stages of body embalming. Mopec™ Left Approach Embalming Sing possesses all the necessary features for full body embalming.
Mopec Left Hand Approach Elevating Wall Mount Autopsy Sink Available on GSA/VA Contract for Federal Government customers only.
Designed to provide an adjustable working area ergonomically correct for users of various heights. Mopec™ Left Hand Approach Elevating Wall Mount Autopsy Sink can be surface mounted or recessed within the thickness of the wall.
  spinner