Autopsy Tables
Mopec Non-Tilting Bariatric Autopsy Cart Available on GSA/VA Contract for Federal Government customers only.
Designed with heavy gauge wall tubing and a reinforced frame with an inset oversized top. Mopec™ Non-Tilting Bariatric Autopsy Cart can be used in conjunction with wall mounted autopsy sinks or as a simple viewing stretcher.
Mopec™ Saddle Autopsy Cart with Various Tray Sizes Available on GSA/VA Contract for Federal Government customers only.
Fitted to saddle a body tray within the frame with rubber bumpers on all four corners. Mopec™ Saddle Autopsy Cart with Various Tray Sizes is a cart that is designed with a built in slope.
Mopec™ Pedestal Style Autopsy Table with Elevating and Bariatric Options Available on GSA/VA Contract for Federal Government customers only.
Pedestal style table that incorporates an optional elevating mechanism within the pedestal. Mopec™ Pedestal Style Autopsy Table with Elevating and Bariatic Options features a hydro-aspirator that is conveniently located near the large sink basin for ease in collection and disposal of fluids.
Mopec DC410 Aluminum False Frame Available on GSA/VA Contract for Federal Government customers only.
Mopec™ DC410 Aluminum False Frame is and optional cover that can be used with Mopec™ Non-Tilting Bariatric Autopsy Cart.
Mopec DA Series Autopsy Cart with Chassis Only Available on GSA/VA Contract for Federal Government customers only.
Possesses the capability of latching ontp a C Series link with its downward sloped edge. Mopec™ DA Series Autopsy Cart with Chassis Only has a rigid frame with a built in slope allowing for proper fluid drainage into the autopsy sink.
Mopec Hand Crank Tilting Autopsy Cart with Plastic X-Ray Top Available on GSA/VA Contract for Federal Government customers only.
Designed with a hand lifting mechanism that elevates one end of the autopsy cart allowing for a sloped surface. Mopec™ Hand Crank Tilting Autopsy Cart with Plastic X-Ray Top can be used in conjunction with any of Mopec™ C Series Wall Mounted Autopsy Sinks.
Mopec Non-Elevating Autopsy Table with Integral Left Wing Available on GSA/VA Contract for Federal Government customers only.
Includes all of the features necessary to perform an autopsy along with an added integrally constructed wing. Mopec™ Non-Elevating Autopsy Table with Integral Left Wing is an L-shaped table with pedestal design that includes standard downdraft ventilation system.
Mopec DE000 Roller Autopsy Cart Available on GSA/VA Contract for Federal Government customers only.
Gliding on corrosion proof nylon rollers, the body tray easily rolls onto the cart frame. Mopec™ DE000 Roller Autopsy Cart is designed with a built in slope and can be used in conjunction with any Mopec™ wall mounted sinks.
Mopec Fold Up Wall Mount Table with Hand Crank Available on GSA/VA Contract for Federal Government customers only.
Mopec™ Fold Up Wall Mount Table provides a compact work area for prep, surgery, and exams.
Mopec Non-Elevating Rotating Autopsy Table CE Series Available on GSA/VA Contract for Federal Government customers only.
Pedestal style table that incorporates a rotating mechanism within the pedestal. Mopec™ Non-Elevating Rotating Autopsy Table CE Series has a downdraft ventilation system which helps ventilate the body during an autopsy.
Mopec DA Series Autopsy Cart with Standard Stainless Steel Top Available on GSA/VA Contract for Federal Government customers only.
Possesses the capability of latching ontp a C Series link with its downward sloped edge. Mopec™ DA Series Autopsy Cart with Standard Stainless Steel Top has a rigid frame with a built in slope allowing for proper fluid drainage into the autopsy sink.
Mopec Hand Crank Tilting Autopsy Cart with Standard Stainless Steel Top Available on GSA/VA Contract for Federal Government customers only.
Designed with a hand lifting mechanism that elevates one end of the autopsy cart allowing for a sloped surface. Mopec™ Hand Crank Tilting Autopsy Cart with Standard Stainless Steel Top can be used in conjunction with any of Mopec™ C Series Wall Mounted Autopsy Sinks.
Mopec Hydraulic Dissection Cart with Chassis Only Available on GSA/VA Contract for Federal Government customers only.
Allow for simple elevating and tilting from either side with dual side mounted foot controls. Mopec™ Hydraulic Dissection Cart with Chassis Only incorporates hydraulic lifting cylinders allowing for either end to be elevated and tilted.
Mopec 78 Inch Pedestal Style Autopsy Table Available on GSA/VA Contract for Federal Government customers only.
Includes all of the features necessary to efficiently perform an autopsy. Mopec™ 78 Inch Pedestal Style Autopsy Table is a pedestal designed table that includes standard downdraft ventilation system and is ideal for pediatric autopsies.
Mopec DA Series Autopsy Cart with Fiberglass X-Ray Top Available on GSA/VA Contract for Federal Government customers only.
Possesses the capability of latching ontp a C Series link with its downward sloped edge. Mopec™ DA Series Autopsy Cart with Fiberglass X-Ray Top has a rigid frame with a built in slope allowing for proper fluid drainage into the autopsy sink.
Mopec GA100 Stainless Steel Autopsy Cart Top Available on GSA/VA Contract for Federal Government customers only.
Mopec™ GA100 Stainless Steel Autopsy Cart Top is ideal for autopsy use and suitable for use with various Mopec™ manufactured carts.
Mopec Elevating Autopsy Table with Integral Left-Handed Wing Available on GSA/VA Contract for Federal Government customers only.
Includes all of the features necessary to perform an autopsy along with an added integrally constructed wing. Mopec™ Elevating Autopsy Table with Integral Left-Handed Wing is an L-shaped table with pedestal design that includes standard downdraft ventilation system.
Mopec DE050 Roller Autopsy Cart Available on GSA/VA Contract for Federal Government customers only.
Gliding on corrosion proof nylon rollers, the body tray easily rolls onto the cart frame. Mopec™ DE050 Roller Autopsy Cart is designed with a built in slope and can be used in conjunction with any Mopec™ wall mounted sinks.
Mopec Plastic Grey X-Ray Autopsy Cart Available on GSA/VA Contract for Federal Government customers only.
Constructed of vacuum formed ABS plastic, making it an incredibly durable and lightweight body tray that is ideal for any medical examiners office. Mopec™ Plastic Grey X-Ray Autopsy Cart Top is designed to universally interface with any line of autopsy carts and body racking systems.
Mopec Hydraulic Dissection Cart with Inner Trough Stainless Steel Top Available on GSA/VA Contract for Federal Government customers only.
Allow for simple elevating and tilting from either side with dual side mounted foot controls. Mopec™ Hydraulic Dissection Cart with Inner Trough Stainless Steel Top incorporates hydraulic lifting cylinders allowing for either end to be elevated and tilted.
Mopec Hand Crank Tilting Autopsy Cart with Fiberglass Trough Top Available on GSA/VA Contract for Federal Government customers only.
Designed with a hand lifting mechanism that elevates one end of the autopsy cart allowing for a sloped surface. Mopec™ Hand Crank Tilting Autopsy Cart with Fiberglass Trough Top can be used in conjunction with any of Mopec™ C Series Wall Mounted Autopsy Sinks.
Mopec Elevating Autopsy Table with Integral Right-Handed Wing Available on GSA/VA Contract for Federal Government customers only.
Includes all of the features necessary to perform an autopsy along with an added integrally constructed wing. Mopec™ Elevating Autopsy Table with Integral Right-Handed Wing is an L-shaped table with pedestal design that includes standard downdraft ventilation system.
Mopec Hydraulic Dissection Cart with Fiberglass X-Ray Top Available on GSA/VA Contract for Federal Government customers only.
Allow for simple elevating and tilting from either side with dual side mounted foot controls. Mopec™ Hydraulic Dissection Cart with Fiberglass X-Ray Top incorporates hydraulic lifting cylinders allowing for either end to be elevated and tilted.
Mopec Hand Crank Tilting Autopsy Cart with Chassis Only Available on GSA/VA Contract for Federal Government customers only.
Designed with a hand lifting mechanism that elevates one end of the autopsy cart allowing for a sloped surface. Mopec™ Hand Crank Tilting Autopsy Cart with Chassis Only can be used in conjunction with any of Mopec™ C Series Wall Mounted Autopsy Sinks.
Mopec Hydraulic Dissection Cart with Plastic X-Ray Top Available on GSA/VA Contract for Federal Government customers only.
Allow for simple elevating and tilting from either side with dual side mounted foot controls. Mopec™ Hydraulic Dissection Cart with Plastic X-Ray Top incorporates hydraulic lifting cylinders allowing for either end to be elevated and tilted.
Mopec Elevating Rotating Autopsy Table CE Series Available on GSA/VA Contract for Federal Government customers only.
Pedestal style table that incorporates a rotating and elevating mechanism within the pedestal. Mopec™ Elevating Rotating Autopsy Table CE Series has a downdraft ventilation system which helps ventilate the body during an autopsy.
Mopec DA Series Autopsy Cart with Inner Trough Stainless Steel Top Available on GSA/VA Contract for Federal Government customers only.
Possesses the capability of latching ontp a C Series link with its downward sloped edge. Mopec™ DA Series Autopsy Cart with Inner Trough Stainless Steel Top has a rigid frame with a built in slope allowing for proper fluid drainage into the autopsy sink.
Mopec Hydraulic Dissection Cart with Standard Stainless Steel Top Available on GSA/VA Contract for Federal Government customers only.
Allow for simple elevating and tilting from either side with dual side mounted foot controls. Mopec™ Hydraulic Dissection Cart with Standard Stainless Steel Top incorporates hydraulic lifting cylinders allowing for either end to be elevated and tilted.
Mopec Hand Crank Tilting Autopsy Cart with Stainless Steel Trough Top Available on GSA/VA Contract for Federal Government customers only.
Designed with a hand lifting mechanism that elevates one end of the autopsy cart allowing for a sloped surface. Mopec™ Hand Crank Tilting Autopsy Cart with Stainless Steel Trough Top can be used in conjunction with any of Mopec™ C Series Wall Mounted Autopsy Sinks.
  spinner