Mortuary Racks
Mopec™ IB Series Portable Cantilever Storage Rack Available on GSA/VA Contract for Federal Government customers only.
A versatile economical means of vertical storage of multiple cadavers that adjusts easily for a wide variety of heights. Mopec™ IB Series Portable Cantilever Storage Rack is an adjustable cantilever racks that is designed to support up to 500 pounds per tier.
Mopec Portable Shelving System Available on GSA/VA Contract for Federal Government customers only.
Eliminates the need to transfer trays and samples during intra-facility transport. Mopec™ Portable Shelving System conveniently stores within the LD500 cabinet for proper ventilation of its contents.
Mopec™ IA Series Stationary Cantilever Storage Rack Available on GSA/VA Contract for Federal Government customers only.
Adjustable cantilever storage racks that are designed to support up to 500 pounds per tier. Mopec™ IA Series Stationary Cantilever Storage Rack is used along with body trays or autopsy cart tops, which can be placed on the rack with a fork type lift.
Mopec Replacement Shelf for Ventilated Tissue Storage System Available on GSA/VA Contract for Federal Government customers only.
Mopec™ Replacement Shelves for Ventilated Tissue Storage System is designed to be able to be arranged to different heights.
Mopec Ventilated Tissue Storage System Available on GSA/VA Contract for Federal Government customers only.
Provides a safe and functional method for storing tissue samples. Mopec™ Ventilated Tissue Storage System is the practical answer to everyday tissue storage and transfer needs.
  spinner