Dissection Saws
Thermo Scientific™ Shandon™ Dual Blade Manual Bone Saw Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Shandon Dual Blade Manual Bone Saw makes simultaneous parallel cuts to remove a slice of bone for processing.
Thermo Scientific™ Shandon™ Dual Blade Manual Bone Saw
Easily cut a wide range of bones, including difficult femoral heads, with this dual blade manual bone saw.
Thermo Scientific™ Shandon™ 10000 Autopsy Saw Available on GSA/VA Contract for Federal Government customers only.
Cut safely and reduce operator fatigue with Shandon™ 10000 Autopsy Saw.
Thermo Scientific™ Shandon™ Stryker™ Autopsy Saw Available on GSA/VA Contract for Federal Government customers only.
Cut quickly and minimize operator fatigue with the easy to handle Stryker™ Autopsy Saw.
Integra™ Miltex™ Satterlee Bone Saw
Light metal handle with stainless steel 8.5 in. (21cm) blade
Integra™ Miltex™ Gigli Saw Handle
Turnable, T-shaped handle
Integra™ Miltex™ MeisterHand™ Gigli Saw Handle
Standard pattern, T-shaped, turnable
Integra™ Miltex™ Gigli Saw
Standard twisted wire type
Mopec 5000 Autopsy Saw
The Mopec 5000 is the first bone saw developed specifically for autopsy use. It is totally submersible for complete cleaning. The handpiece runs on 40V for a cooler touch, reduced vibration; and the axial mount blades offer safer installation.
Mopec Autopsy Saw
This Mopec saw has a rugged construction and a machined aluminum housing that reduces heat and noise. On/Off switch is designed and placed so that it will not break if dropped. Designed for removing the cranial cap, making linear cuts or sectioning small bone specimens.
  spinner