Saw Blades
Thermo Scientific™ Shandon™ Large Section Blades
Remove skull cap or bone specimens with general purpose Thermo Scientific™ Shandon™ Large Section Blades.
Thermo Scientific™ Shandon™ Round Autopsy Saw Blades
Cut to a depth of 1.75″ (44mm) with these round autopsy saw blades.
Thermo Scientific™ Shandon™ Spinal Column Saw Blade
Remove one square-inch (645 square mm) of bone with this specialized spinal column saw blade.
Thermo Scientific™ Shandon™ Extra Arbor, Arbor
Use Thermo Scientific™ Shandon™ Extra Arbor with all Shandon autopsy blades.
Mopec Autopsy Saw Blades, Compatible with Stryker Saw
These high quality affordable autopsy saw blades from Mopec are hardened to prolong life and sharpness.
Mopec Autopsy Saw Blades for Mopec 5000
Round, sectional and segmented blades from Mopec are available for any bone cutting application including cranial removals, cross and long bone sectioning and vertebral column. Axial mount blades eliminate nicks to fingers and knuckles.
Mopec Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw
Mopec's exclusive titanium coating assures longer and sharper blade life. Fits all saws with radial mount arbors.
  spinner